Justrite Oily Waste Can
Prevent fires or spontaneous combustion
  • Elevated base reduces heat, prevents moisture build up
  • FM Approved
  • OSHA compliant
  • Certified by UL/ULC, TÜV
22,500.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Capacity (Liters) Material
Availability Pricing
Justrite 09100 Oily Waste Can, Steel,  6 Gal 8628432 Mfr # 09100
COMPARE
6 22.71 Steel
₹22,500.00INR / Each