Julabo MAGIO™ MS Refrigerated/Heating Circulators
Extremely powerful pressure/suction pump perfect for external temperature tasks
  • Performance values up to 31 L/min or 0.92 bar (pressure)/-0.4 bar (suction)
  • All wetted parts are constructed of stainless steel for chemical resistance
  • High-resolution, TFT touch-screen display features a multi-lingual user interface
  • Remote control and data management made easy with USB interface, RS-232, and Ethernet
  • Pt100 external sensor connection for measurement and control
The MAGIO™ refrigerated/heating circulators stand out thanks to their premium quality, high performance, and intuitive operation. The units offer extra strong pressure and suction pumps, meeting the highest demands for temperature control of external applications. Whether in basic research, material testing or technical systems, the MAGIO™ refrigerated circulators offer high-tech solutions for high customer requirements.
2,760,000.00 - ₹5,123,000.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 20° C (watts) Min Temperature (° C) Power (VAC)
Availability Pricing
Julabo MAGIO™ MS-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 1 kW Heater and Cooling 330 W at 20 °C, R449A; 115 VAC, 60 Hz WW-12153-00 Mfr # 9032713.S1.02
COMPARE
330 -30 115
₹2,760,000.00INR / Each
Julabo MAGIO™ MS-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 2 kW Heater and Cooling 330 W at 20 °C, R449A; 200-230 VAC, 50/60 Hz WW-12153-01 Mfr # 9032713.S1.33
COMPARE
330 -30 200 / 230
₹2,760,000.00INR / Each
Julabo MAGIO™ MS-600F Refrigerated/Heating Circulator, 7.5 L Tank, -35 to +200 °C, 1 kW Heater and Cooling 600 W at 20 °C, R449A; 115 VAC, 60 Hz WW-12153-02 Mfr # 9032704.02
COMPARE
600 -35 115
₹3,229,000.00INR / Each
Julabo MAGIO™ MS-600F Refrigerated/Heating Circulator, 7.5 L Tank, -35 to +200 °C, 2 kW Heater and Cooling 600 W at 20 °C, R449A; 200-230 VAC, 50/60 Hz WW-12153-03 Mfr # 9032704.33
COMPARE
600 -35 200 / 230
₹3,229,000.00INR / Each
Julabo MAGIO™ MS-601F Refrigerated/Heating Circulator, 10 L Tank, -35 to +200 °C, 1 kW Heater and Cooling 600 W at 20 °C, R449A; 115 VAC, 60 Hz WW-12153-04 Mfr # 9032705.02
COMPARE
600 -35 115
₹3,382,000.00INR / Each
Julabo MAGIO™ MS-601F Refrigerated/Heating Circulator, 10 L Tank, -35 to +200 °C, 2 kW Heater and Cooling 600 W at 20 °C, R449A; 200-230 VAC, 50/60 Hz WW-12153-05 Mfr # 9032705.33
COMPARE
600 -35 200 / 230
₹3,382,000.00INR / Each
Julabo MAGIO™ MS-1000F Refrigerated/Heating Circulator, 7.5 L Tank, -50 to +200 °C, 1 kW Heater and Cooling 1000 W at 20 °C, R449A; 115 VAC, 60 Hz WW-12153-08 Mfr # 9032707.02
COMPARE
1000 -50 115
₹4,347,000.00INR / Each
Julabo MAGIO™ MS-1000F Refrigerated/Heating Circulator, 7.5 L Tank, -50 to +200 °C, 2 kW Heater and Cooling 1000 W at 20 °C, R449A; 200-230 VAC, 50/60 Hz WW-12153-09 Mfr # 9032707.33
COMPARE
1000 -50 200 / 230
₹4,347,000.00INR / Each
Julabo MAGIO™ MS-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 1 kW Heater and Cooling 330 W at 20 °C, R290; 115 VAC, 60 Hz WW-12153-66 Mfr # 9032713.N1.02
COMPARE
330 -30 115
₹2,760,000.00INR / Each
Julabo MAGIO™ MS-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 2 kW Heater and Cooling 330 W at 20 °C, R290; 200-230 VAC, 50/60 Hz WW-12153-67 Mfr # 9032713.N1.33
COMPARE
330 -30 200 / 230
₹2,760,000.00INR / Each
Julabo MAGIO™ MS-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 1 kW Heater and Cooling 460 W at 20 °C, R290; 115 VAC, 60 Hz WW-12153-68 Mfr # 9032714.N1.02
COMPARE
460 -30 115
₹2,959,000.00INR / Each
Julabo MAGIO™ MS-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 2 kW Heater and Cooling 460 W at 20 °C, R290; 200-230 VAC, 50/60 Hz WW-12153-69 Mfr # 9032714.N1.33
COMPARE
460 -30 200 / 230
₹2,959,000.00INR / Each
Julabo MAGIO™ MS-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 1 kW Heater and Cooling 460 W at 20 °C, R449A; 115 VAC, 60 Hz WW-12153-70 Mfr # 9032714.S1.02
COMPARE
460 -30 115
₹2,959,000.00INR / Each
Julabo MAGIO™ MS-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, 2 kW Heater and Cooling 460 W at 20 °C, R449A; 200-230 VAC, 50/60 Hz WW-12153-71 Mfr # 9032714.S1.33
COMPARE
460 -30 200 / 230
₹2,959,000.00INR / Each
Julabo MAGIO™ MS-1000FW Refrigerated/Heating Circulator, 7.5 L Tank, -50 to +200 °C, 1 kW Heater and Cooling 1000 W at 20 °C, R449A; 115 VAC, 60 Hz WW-12153-72 Mfr # 9032727.02
COMPARE
1000 -50 115
₹5,123,000.00INR / Each
Julabo MAGIO™ MS-1000FW Refrigerated/Heating Circulator, 7.5 L Tank, -50 to +200 °C, 1 kW Heater and Cooling 1000 W at 20 °C, R449A; 200-230 VAC, 50/60 Hz WW-12153-73 Mfr # 9032727.33
COMPARE
1000 -50 200 / 230
₹5,123,000.00INR / Each
Showing 1- 16 of 16