Julabo FL and FLW Series Recirculating Chillers
Recirculating coolers for environmentally friendly chilling
  • Access the power and temperature controls, RS-232 port, and drain—all from the front
  • Splashproof, large-digit, LED display shows temperature and fill-level
  • Easily fill unit from the top—includes protective lid
  • Low-level audible and optical alarms protect your process and the unit
  • Accurate PID temperature control with 0.1°C resolution
A wide variety of cooling capacities make these recirculators ideal for laboratory and general-use applications. PID temperature controller with LED display includes single-point temperature calibration for maximum accuracy. Powerful immersion pumps make them suitable for continuous operation, and RS-232 interface provides temperature data transfer to a PC.
These recirculating chillers are ergonomically designed for easy operation. Features include top filling with digital fill-level indicator; accessible drain tap; no side vents—place next to other instruments; and a removable grid for condenser cleaning to keep your chiller running at full cooling capacity. Environment-friendly operation with low energy consumption.
1,763,000.00 - ₹3,257,330.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 20° C (watts) Power (VAC) Description
Availability Pricing
Julabo FL300   Recirculating Chiller, 300 W, -20 to +40°C; 115V/60Hz WW-12150-21 Mfr # 9660003.2
COMPARE
300 115 Recirculating Chiller, 300 W, -20 to +40°C; 115V/60Hz
₹1,763,000.00INR / Each
Julabo FL300   Recirculating Chiller, 300 W, -20 to +40°C; 220V/60Hz WW-12150-22 Mfr # 9660003.13
COMPARE
300 220 Recirculating Chiller, 300 W, -20 to +40°C; 220V/60Hz
₹1,763,000.00INR / Each
Julabo FL1201 Recirculating Chiller, 1200 W, -20 to +40°C; 115V/60Hz WW-12150-25 Mfr # 9661012.2
COMPARE
1200 115 Recirculating Chiller, 1200 W, -20 to +40°C; 115V/60Hz
₹2,532,000.00INR / Each
Julabo FL1701 Recirculating Chiller, 1700 W, -20 to +40°C; 115V/60Hz WW-12150-29 Mfr # 9661017.2
COMPARE
1700 115 Recirculating Chiller, 1700 W, -20 to +40°C; 115V/60Hz
₹2,713,000.00INR / Each
Julabo FL1703 Recirculating Chiller, 1700 W, -20 to +40°C, 230V/60Hz WW-12150-31 Mfr # 9663017.13
COMPARE
1700 230 Recirculating Chiller, 1700 W, -20 to +40°C; 230V/60Hz
₹3,086,000.00INR / Each
Julabo FL1703 Recirculating Chiller, 1700 W, -20 to +40 °C; 230V/50Hz, UK Plug WW-12154-24 Mfr # 9663017.04
COMPARE
1700 230 Recirculating Chiller, 1700 W, -20 to +40 °C; 230V/50Hz, UK
₹3,257,330.00INR / Each