Julabo DYNEO™ DD Refrigerated/Heating Circulators
Ensures rapid heat-up and cool-down times
  • Easily switch between internal and external applications
  • Central rotary knob simplifies operation
  • Clearly view menu options from large TFT color display
  • Built-in drain tap for easy and safe drainage
  • Meets Class III (FL) according to DIN 12876-1
These DYNEO™ DD refrigerated/heating circulators, equipped with thermal insulated stainless steel closed bath tanks, can easily be switched between internal and external circulation. The adjustable pressure pump has a flow rate of 27 L/min and a flow pressure of 10.2 psi. The tanks include a space-saving cooling coil and drain tap for safe and easy removal of liquids.
The control panel features a multilingual 3.5” TFT color display with a central rotary control knob for straightforward operation. Other features include an integrated programmer, external Pt100 sensor connection, pump connectors (M16 x 1, male), automatic high-temperature shutoff, low liquid level cutoff, and ergonomic handles.
All models are equipped with USB interface. Circulators are available in a wide range of working temperatures and can be purchased with an optional RS-232 or analog interface.
2,634,000.00 - ₹4,734,000.00INR / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Interface Power (VAC) Reservoir Capacity (L)
Availability Pricing
Julabo DYNEO™ DD-200F Refrigerated/Heating Circulator, 4 L Tank, -20 to +200 °C, USB and RS-232 Interface, R134a; 115 VAC, 60 Hz 1215234 Mfr # 9021701.02.D
COMPARE
USB, RS-232 115 4
₹2,778,000.00INR / Each
Julabo DYNEO™ DD-600F Refrigerated/Heating Circulator, 7.5 L Tank, -35 to +200 °C, USB Interface, R449A; 115 VAC, 60 Hz 1215250 Mfr # 9021704.02
COMPARE
USB 115 7.5
₹3,287,000.00INR / Each
Julabo DYNEO™ DD-600F Refrigerated/Heating Circulator, 7.5 L Tank, -35 to +200 °C, USB and RS-232 Interface, R449A; 115 VAC, 60 Hz 1215252 Mfr # 9021704.02.D
COMPARE
USB, RS-232 115 7.5
₹3,406,000.00INR / Each
Julabo DYNEO™ DD-1000F Refrigerated/Heating Circulator, 7.5 L Tank, -50 to +200 °C, USB and RS-232 Interface, R449A; 115 VAC, 60 Hz 1215270 Mfr # 9021707.02.D
COMPARE
USB, RS-232 115 7.5
₹4,734,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB Interface, R290; 115 VAC, 60 Hz 1215336 Mfr # 9021713.N1.02
COMPARE
USB 115 4
₹2,852,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and Analog Interface, R290; 115 VAC, 60 Hz 1215337 Mfr # 9021713.N1.02.A
COMPARE
USB, Analog 115 4
₹3,081,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and RS-232 Interface, R290; 115 VAC, 60 Hz 1215338 Mfr # 9021713.N1.02.D
COMPARE
USB, RS-232 115 4
₹2,984,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB Interface, R449A; 115 VAC, 60 Hz 1215339 Mfr # 9021713.S1.02
COMPARE
USB 115 4
₹2,634,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and Analog Interface, R449A; 115 VAC, 60 Hz 1215340 Mfr # 9021713.S1.02.A
COMPARE
USB, Analog 115 4
₹2,844,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and RS-232 Interface, R449A; 115 VAC, 60 Hz 1215341 Mfr # 9021713.S1.02.D
COMPARE
USB, RS-232 115 4
₹2,766,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB Interface, R290; 200-230 VAC, 50/60 Hz 1215342 Mfr # 9021713.N1.33
COMPARE
USB 230 4
₹2,852,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and Analog Interface, R290; 200-230 VAC, 50/60 Hz 1215343 Mfr # 9021713.N1.33.A
COMPARE
USB, Analog 230 4
₹3,081,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and RS-232 Interface, R290; 200-230 VAC, 50/60 Hz 1215344 Mfr # 9021713.N1.33.D
COMPARE
USB, RS-232 230 4
₹2,984,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB Interface, R449A; 200-230 VAC, 50/60 Hz 1215345 Mfr # 9021713.S1.33
COMPARE
USB 230 4
₹2,634,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and Analog Interface, R449A; 200-230 VAC, 50/60 Hz 1215346 Mfr # 9021713.S1.33.A
COMPARE
USB, Analog 230 4
₹2,844,000.00INR / Each
Julabo DYNEO™ DD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and RS-232 Interface, R449A; 200-230 VAC, 50/60 Hz 1215347 Mfr # 9021713.S1.33.D
COMPARE
USB, RS-232 230 4
₹2,766,000.00INR / Each
Julabo DYNEO™ DD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB Interface, R290; 115 VAC, 60 Hz 1215348 Mfr # 9021714.N1.02
COMPARE
USB 115 4
₹2,867,000.00INR / Each
Julabo DYNEO™ DD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB Interface, R290; 200-230 VAC, 50/60 Hz 1215349 Mfr # 9021714.N1.33
COMPARE
USB 230 4
₹3,100,000.00INR / Each
Julabo DYNEO™ DD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and Analog Interface, R290; 115 VAC, 60 Hz 1215350 Mfr # 9021714.N1.02.A
COMPARE
USB, Analog 115 4
₹3,078,000.00INR / Each
Julabo DYNEO™ DD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to +200 °C, USB and Analog Interface, R290; 200-230 VAC, 50/60 Hz 1215351 Mfr # 9021714.N1.33.A
COMPARE
USB, Analog 230 4
₹3,327,000.00INR / Each
Showing 1 - 20 of 28