Jeio Tech Flask Support, Polyproplylene
Compact design conserves valuable laboratory space
  • Energy-saving system performs tests efficiently, reducing power consumption during operation
  • Quickly check operation status with 5.6” color touch display controller and operation status LED lights
  • Double-sealed door provides enhanced airtightness and efficiency
  • Internal chamber constructed of stainless steel for easy cleaning and excellent corrosion resistance
  • Easily observe samples with tempered glass door with built-in heating wire and LED lamp
These Lab Companion TC3 low-temperature environmental chambers feature excellent thermal insulation and double-sealing door structure ensure temperature reproducibility even for repeated test use. Easily control and view operations with the 5.6” color touch display controller. Operation status is also displayed with LED colors, convenient to identify chamber status from a far.
Chambers provide enhanced safety with multiple protections and alarms. Chambers feature electronic over temperature shutdown, door open warning and shut off, overcurrent and short protection, compressor over temperature, and high/low refrigerant shut off.
4,074,000.00 - ₹6,880,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Min Temperature (° C) Description
Availability Pricing
Jeio Tech TC3-ME-025 Low-Temperature Chamber, 25 L, -20 to 150°C; 230V 50/60 Hz 5240307 Mfr # AAH860132K
COMPARE
0.88 -20 Low-Temperature Chamber, 25 L, -20 to 150°C; 230V 50/60 Hz
₹4,074,000.00INR / Each
Jeio Tech TC3-ME-065 Low-Temperature Chamber, 65 L, -20 to 150°C; 230V 50/60 Hz 5240308 Mfr # AAH860232K
COMPARE
2.3 -20 Low-Temperature Chamber, 65 L, -20 to 150°C; 230V 50/60 Hz
₹4,911,000.00INR / Each
Jeio Tech TC3-ME-100 Low-Temperature Chamber, 100 L, -20 to 150°C; 230V 50/60 Hz 5240309 Mfr # AAH860332K
COMPARE
3.553 -20 Low-Temperature Chamber, 100 L, -20 to 150°C; 230V 50/60 Hz
₹5,175,000.00INR / Each
Jeio Tech TC3-KE-025 Table Top Low-Temperature Chamber, 25 L, -40 to 150°C; 230V 60 Hz 5240312 Mfr # AAH861131K
COMPARE
0.88 -40 Table Top Low-Temperature Chamber, 25 L, -40 to 150°C; 230V 60 Hz
₹5,420,000.00INR / Each
Jeio Tech TC3-KE-025 Table Top Low-Temperature Chamber, 25 L, -40 to 150°C; 230V 50 Hz 5240313 Mfr # AAH861132K
COMPARE
0.88 -40 Table Top Low-Temperature Chamber, 25 L, -40 to 150°C; 230V 50 Hz
₹5,420,000.00INR / Each
Jeio Tech TC3-KE-065 Table Top Low-Temperature Chamber, 65 L, -40 to 150°C; 230V 60 Hz 5240314 Mfr # AAH861231K
COMPARE
2.3 -40 Table Top Low-Temperature Chamber, 65 L, -40 to 150°C; 230V 60 Hz
₹6,324,000.00INR / Each
Jeio Tech TC3-KE-065 Table Top Low-Temperature Chamber, 65 L, -40 to 150°C; 230V 50 Hz 5240317 Mfr # AAH861232K
COMPARE
2.3 -40 Table Top Low-Temperature Chamber, 65 L, -40 to 150°C; 230V 50 Hz
₹6,324,000.00INR / Each
Jeio Tech TC3-KE-100 Table Top Low-Temperature Chamber, 100 L, -40 to 150°C; 230V 60 Hz 5240318 Mfr # AAH861331K
COMPARE
3.553 -40 Table Top Low-Temperature Chamber, 100 L, -40 to 150°C; 230V 60 Hz
₹6,880,000.00INR / Each
Jeio Tech TC3-KE-100 Table Top Low-Temperature Chamber, 100 L, -40 to 150°C; 230V 50 Hz 5240319 Mfr # AAH861332K
COMPARE
3.553 -40 Table Top Low-Temperature Chamber, 100 L, -40 to 150°C; 230V 50 Hz
₹6,838,000.00INR / Each
Showing 1- 9 of 9