Intellitech i-FILL® Micro Pump Accessories
Essential replacement parts and disposable fluid paths engineered to maintain the accuracy, reliability, and hygienic performance of Intellitech i-FILL Micro liquid filling systems
  • Genuine Intellitech components designed for full compatibility with i-FILL micro pumps
  • Disposable fluid path kits minimize cross-contamination and eliminate cleaning downtime
  • Precision-machined pistons ensure consistent, repeatable dispense volumes
  • Ideal for laboratory, pharmaceutical, and R&D liquid filling applications
These Intellitech replacement parts and consumables, including disposable fluid path kits, precision pistons, and tubing components, are engineered to support the long-term performance of the i-FILL Micro benchtop liquid filling system. Each component is manufactured to exact specifications to ensure reliable operation, accurate dispensing, and seamless integration with existing equipment.
Disposable fluid path kits are gamma-irradiated and single-use, reducing the risk of cross-contamination while streamlining changeovers between products or batches. Replacement pistons and tubing components restore optimal pump performance and are ideal for routine maintenance, validation, or backup inventory. Together, these components help maintain uptime, improve process consistency, and support regulated laboratory and production environments.
18,900.00 - ₹42,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Intellitech i-FILL® 1 mL Kit For Micro Pump 9140082 Mfr # M2-00445
COMPARE
1 mL Kit For Micro Pump
₹18,900.00INR / Each
Intellitech i-FILL® 10 mL Kit For Micro Pump 9140083 Mfr # M2-00457
COMPARE
10 mL Kit For Micro Pump
₹18,900.00INR / Each
Intellitech i-FILL® Stand For Micro Pump 9140086 Mfr # M1-01975
COMPARE
Stand For Micro Pump
₹28,000.00INR / Each
Intellitech i-FILL® Foot Switch For Micro Pump 9140087 Mfr # T-91-SWANOF
COMPARE
Foot Switch For Micro Pump
₹42,000.00INR / Each