Intelligent Digital Sensors (IDS) for WTW MultiLine Meters

137,000.00 - ₹336,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Accessory Type Range Description
Availability Pricing
WTW SenTix 940 IDS pH Electrode, Non-refillable; 1.5 m Cable 9957530 Mfr # 103740
COMPARE
pH Electrode 0 to 14 IDS pH Electrode, Non-refillable; 1.5 m Cable
₹137,000.00INR / Each
WTW TetraCon 925 IDS Conductivity Probe, 2000 mS; 1.5 m Cable 9957537 Mfr # 301710
COMPARE
Conductivity Cell 10 µS to 2000 mS at ±0.5 % of value IDS Conductivity Probe, 2000 mS; 1.5 m Cable
₹245,000.00INR / Each
WTW TetraCon 925-3 IDS Conductivity Probe, 2000 mS; 3 m Cable 9957538 Mfr # 301711
COMPARE
Conductivity Cell 10 µS to 2000 mS at ±0.5 % of value IDS Conductivity Probe, 2000 mS; 3 m Cable
₹251,000.00INR / Each
WTW LR 925/01 IDS Conductivity Probe, 200 uS; 1.5 m Cable 9957541 Mfr # 301720
COMPARE
Conductivity Cell 0.01 to 200 µS at ±0.5 % of value IDS Conductivity Probe, 200 uS; 1.5 m Cable
₹336,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat