InnoCal® NIST-traceable calibrations for electrical transmitters
NIST-traceable calibrations for electrical transmitters
  • Includes a NIST-traceable calibration certificate that establishes traceability to NIST
  • Certificate lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
29,800.00 - ₹41,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Description
Availability Pricing
COMPARE
Electrical NIST-Traceable Calibration; AC/DC Current/Voltage Transmitter
₹29,800.00INR / Each
COMPARE
A2LA ISO17025 Accredited Electrical A2LA ISO17025 Accredited Calibration, AC/DC Current/Voltage Transmitter
₹41,900.00INR / Each