Imada Digital Force Gauge with USB Output
  • Processes 1,000 data values/second and transmits all of them, not just the peak
  • Programmable high/low setpoints with color-coded LED indicators for Go/No Go testing
  • lbf, Kgf, and N units are selectable
  • 1,000 Data memory for computer download
  • USB, RS-232C, and Analog outputs
  • Ergonomic design with non-slip rubber grip, heavy duty metal construction
  • Overload capacity 200% of F.S.
  • Real time and peak mode (selectable)
  • Tare function
  • Reversible display
  • Full set of attachments, ZP Logger (basic data acquisition software), AC adapter/charger, carrying case and Certificate of Calibration traceable to NIST included
ZP digital force gauges are capable of processing 1,000 data/second. Collect peak data on your computer with the USB cable and ZP Logger (software included).
345,000.00 - ₹381,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Imada ZTA-11 Force Gauge with USB output, 11 lb capacity 5985014 Mfr # ZTA-11
COMPARE
Capacity: 11 lb/5 kg/50 N
₹345,000.00INR / Each
Imada ZTA-110 Gauge W/Usb Output 110 LBF Cap. 5985016 Mfr # ZTA-110
COMPARE
Capacity: 110 lb/50 kg/500 N
₹345,000.00INR / Each
Imada ZTA-220 Gauge Usb Output 220 LBF Cap. 5985017 Mfr # ZTA-220
COMPARE
Capacity: 220 lb/100 kg/1000 N
₹381,000.00INR / Each