IKA Vortex 3 Shaker
Wide speed range and variety of attachments make this ideal for any lab
  • Adjustable speed control from 250 to 2500 rpm
Compact, versatile shaker offers short-time, touch-activated function or continuous operation. Variable speed control and optional attachments make this shaker ideal for a wide range of mixing applications. Silicone feet and ultra-high vibration dampening base make this shaker stable at high speeds. Self-ventilated motor provides for continuous operation with minimal heating. All accessories (sold separately) click onto unit in virtually any position.
42,700.00 - ₹200,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
IKA Vortex 3 Shaker, 0 to 2500 rpm, 115 VAC 0430410 Mfr # 3340001
COMPARE
115
₹200,000.00INR / Each
IKA Vortex 3 Shaker, 0 to 2500 rpm, 230 VAC 0430412 Mfr # 0003340000
COMPARE
230
₹42,700.00INR / Each