IKA Topolino Magnetic Stirrer
Mini Stirrer saves valuable benchtop space
  • Continually adjustable speed range up to 1800 rpm
  • Durable polypropylene top plate
This compact, low-profile mini-stirrer is perfect for mixing quantities up to 250 mL. Its small size makes it convenient for use anywhere in the lab and maximizes premium benchtop space. Features a dual-voltage, durable brushless motor and high magnetic adhesion.
10,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
IKA Topolino Compact Magnetic Stirrer; 115/230 VAC 8400213 Mfr # 0003368000
COMPARE
115 / 230
₹10,100.00INR / Each