IKA® T25 Easy Clean Homogenizers
Quick and easy cleaning of the rotor-stator generators—no disassembling required
  • Rotor-stator generators cleaning status allows for quick cleaning saving on down time
  • LED speed and timer display
  • USB interface for control and documentation via labworldsoft®
  • Increased safety through tool recognition and IP30 rating
  • Choose from systems with or without temperature measurement
The T25 easy clean system is ideal for batch homogenizing of cell tissues, emulsifying suspended solids, and air pollution filter extraction. The compatible rotor-stator generators (order rotor-stator generators separately) feature cleaning status indicator to inform you of when cleaning is required. Cleaning is quick and easy—no disassembling required. Shielded in a 316 stainless steel shaft, the rotor acts as a centrifugal pump to recirculate the liquid and suspended solids through the generator, where shear, impact, collision, and cavitation provide rapid homogenization.
The T25 Control homogenizer along with the rotor stator generator allows continuous temperature measurement with an integrated temperature sensor. Device overheating is prevented by continuous temperature monitoring and the setting of temperature limits.
301,112.00 - ₹370,854.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
IKA T 25 easy clean Digital Disperser; 1/Ea 1635343 Mfr # 25002582
COMPARE
₹301,112.00INR / Each
IKA T25 easy clean control Disperser; 1/Ea 1635484 Mfr # 25002522
COMPARE
₹370,854.00INR / Each