IKA® T25 easy clean digital Homogenizers
Quick and easy cleaning of the rotor-stator generators—no disassembling required
  • Rotor-stator generators cleaning status allows for quick cleaning saving on down time
  • USB interface for control and documentation via labworldsoft® software
  • Increased safety through tool recognition and IP30 rating
  • Choose from systems with or without temperature measurement
The T25 easy clean system is ideal for batch homogenizing of cell tissues, emulsifying suspended solids, and air pollution filter extraction. The compatible rotor-stator generators (order rotor-stator generators separately) feature cleaning status indicator to inform you of when cleaning is required. Cleaning is quick and easy—no disassembling required. Shielded in a 316 stainless steel shaft, the rotor acts as a centrifugal pump to recirculate the liquid and suspended solids through the generator, where shear, impact, collision, and cavitation provide rapid homogenization.
The T25 easy clean control homogenizer along with the 04739-25 rotor stator generator allows continuous temperature measurement with an integrated temperature sensor. Device overheating is prevented by continuous temperature monitoring and the setting of temperature limits.
840,000.00 - ₹923,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Max Speed (rpm) Description
Availability Pricing
IKA T 25 Easy Clean Homogenizer, Digital, 3,000 to 25,000 rpm; 100-240 VAC 50/60 Hz 0473923 Mfr # 0025002561
COMPARE
2000 25000 Easy Clean Homogenizer, Digital, 3,000 to 25,000 rpm; 100-240 VAC 50/60 Hz
₹840,000.00INR / Each
IKA T 25 Easy Clean Homogenizer, Control, 3,000 to 25,000 rpm; 100-240 VAC 50/60 Hz 0473924 Mfr # 0025002501
COMPARE
2000 25000 Easy Clean Homogenizer, Control, 3,000 to 25,000 rpm; 100-240 VAC 50/60 Hz
₹923,000.00INR / Each