IKA T 10 Basic Ultra Turrax Homogenizer
125 Watt motor processes sample volumes of 0.5 to 100 mL up to 30,000 rpm
  • Light weight and ergonomic form is ideal for manual operation
  • Quick-release coupling simplifies the removal of dispersing elements without the need for tools
  • Available disposable dispersion elements eliminate cross contamination -- great for PCR applications
These compact Homogenizers are great for high-speed hand held operation. The 125-Watt motor offers stable operation in a variety of media samples, and variable speed control from 8,000 to 10,000 rpm. Disposable dispersion elements eliminate concerns of cross-contamination with DNA, or PCR analysis. Stainless steel dispersion elements remove quickly and clean completely for reuse. The rotor-stator generator uses a pump-like action to constantly draw in sample. Through a combination of shear, impact, collision, and cavitation, the generator rapidly disperses and disintegrates the material suspended in the media.
135,000.00 - ₹442,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
IKA 3737001 T10 basic ULTRA-TURRAX Homogenizer system 115VAC 0472050 Mfr # 3737001
COMPARE
50/60 115
₹442,000.00INR / Each
IKA 3737000 T10 basic ULTRA-TURRAX Homogenizer system 230VAC 0472051 Mfr # 3737000
COMPARE
50/60 230
₹135,000.00INR / Each