IKA® RV 8 Rotary Evaporators
Updated design for standard evaporating applications
  • Easy-to-use manual lift with integrated safety lift-out function
  • Ambidextrous design makes operation more comfortable
  • Adjustable immersion angle and height for maximum heating efficiency
  • Compatible with complete range of IKA rotary evaporator glassware
The RV 8 vertical distilling system is designed for ease of use in all standard evaporating applications. The manual lift with ambidextrous design lets you precisely position the glassware. Digital displays indicate both rotation speed and heating bath temperature, allowing for optimal control of the process. Easy-to-use clamp system simplifies changing evaporator flasks.
Ergonomic 4 L water/oil heating bath has a locking function for temperature settings from ambient to 180°C. Bath is designed with integrated ergonomic carrying handles for convenient positioning and oblong design for fast heating.
Systems are available with standard glassware, with dry-ice condenser glassware, or with no glassware. Safety-coated glassware options are available; they are resistant to acids and solvents and protect against shattered glass.
1,340,000.00 - ₹1,638,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bath Type Condenser Type Glass Type
Availability Pricing
IKA RV 8 V Rotary Evaporator with Uncoated Glassware and Digital Speed/Temperature Control; 120 VAC 2871135 Mfr # 0010003570
COMPARE
Water/Oil Vertical Uncoated
₹1,340,000.00INR / Each
IKA RV 8 V-C Rotary Evaporator with Coated Glassware and Digital Speed/Temperature Control; 120 VAC 2871136 Mfr # 0010003571
COMPARE
Water/Oil Vertical Safety-coated
₹1,415,000.00INR / Each
IKA RV 8 Rotary Evaporator with Dry Ice Condenser, Coated Glassware and Digital Speed/Temperature Control; 120 VAC 2871138 Mfr # 0010003574
COMPARE
Water/Oil Cold-trap Safety-coated
₹1,638,000.00INR / Each