IKA RH Basic and Digital Stirring Hot Plates
Stir volumes up to 15 liters
These economical stirring hot plates feature a soft start motor to prevent samples from splashing and spraying. Stirrers produce a strong magnetic field to create speeds that are adjustable from 50 to 2000 rpm. The composite stainless steel plate ensures rapid heat up to the desired temperature up to 608°F (320°C). Produce highly accurate temperature control by connecting stirring hot plates to an external sensor according to DIN 12878. An indicator warns when the plate is hot to prevent burns. Digital model features an easy-to-operate keypad with a digital display of temperature and speed. All models feature a built-in mount for a support rod.
187,000.00 - ₹209,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
IKA 5019701 RH Basic Stirring Hot Plate, 115V (Top plate material: Stainless Steel) 0467112 Mfr # 5019701
COMPARE
115
₹187,000.00INR / Each
IKA 5019801 RH Digital Stirring Hot Plate, 115V (Digital) 0467114 Mfr # 5019801
COMPARE
115
₹209,000.00INR / Each
IKA 5019800 RH Digital Stirring Hot Plate, 230V (Digital) 0467116 Mfr # 0005019800
COMPARE
230
₹209,000.00INR / Each