IKA RET control-visc Digital Stirring Hot Plate with Weighing and pH Functions
First stirring hot plate to weigh, check pH, and monitor temperature
  • Weigh and stir samples in one convenient unit
  • Monitor pH of your sample with built-in BNC connector
  • Built-in temperature sensor for precise temperature control
This innovative unit offers the most advanced features within the market today, providing you the ability to monitor your sample activity more accurately. Monitor and weigh samples up to 5 kg using the integrated weighing function. Built-in BNC connector allows you to attach an electrode for analyzing and adjusting the pH of your sample. Set hot plate temperature up to 644°F (340°C) and observe the actual temperature of the sample with the temperature probe. Easily examine all parameters on the alphanumeric graphical TFT LCD with excellent image quality, easy-to-use navigation control, torque trend, and timer/countdown functions.
Round stainless steel top provides quick and effective heat transfer to your samples. Improved heat control technology increases safety, while hot top indicator warns users to help prevent burns and injuries. Utilize the adjustable, from 122 to 698°F (50 to 370°C), over-temperature safety circuit to protect samples from overheating. Built-in RS-232 interface allows direct connection to a PC for control and documentation of trials for later study.
248,842.00 - ₹613,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Max Stirring Volume (Liters) Power (VAC)
Availability Pricing
IKA 5020001 RET control-visc Stirring Hot Plate, 115V 0467129 Mfr # 5020001
COMPARE
1700 20 115
₹613,000.00INR / Each
IKA 5020000 RET control-visc Stirring Hot Plate, 230V 0467131 Mfr # 0005020000
COMPARE
1700 20 230
₹248,842.00INR / Each