IKA EUROSTAR 40 Digital Mixers
Maintenance-free, continuous-duty mixers ideal for general laboratory use
  • Simple, reliable mixers for volumes up to 6.6 gallons (25 liters) and viscosities up to 30,000 cps
  • Stirring speeds up to 2000 rpm are automatically maintained as viscosity changes
These mixers feature a through-shaft design that permits adjustment of shaft length and impeller depth—allowing you to place or remove mixing vessels without repositioning the mixer. The motor and drive unit are conveniently integrated into a compact aluminum and thermoplastic housing with easy-to-use controls and a clear LED display of rpm. Mixers feature infinitely adjustable stepless speed control. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in fluid viscosity.
Quiet, fan-cooled motors feature thermal overload shut-off, and an efficient heat-dissipating design to ensure a long service life. Motor is maintenance-free and rated for continuous duty. Safety circuits ensure automatic cut-off in an anti-stall or overload condition.
132,239.00 - ₹564,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
IKA Eurostar 40 Digital Mixer, 30 to 2000 rpm, 115 VAC 5070240 Mfr # 4444001
COMPARE
2000 56.6 115
₹564,000.00INR / Each
IKA Eurostar 40 Digital Mixer, 30 to 2000 rpm, 230 VAC 5070245 Mfr # 0004444000
COMPARE
2000 56.6 230
₹132,239.00INR / Each