IKA EUROSTAR 200 Digital Constant-Speed Mixers
Powerful motors handle volumes up to 26.4 gallons (100 liters)
  • Mixers maintain set speed even as fluid viscosity changes
  • Dual-range EUROSTAR 200 Digital will handle fluid viscosities up to 100,000 cps
These mixers feature a through-shaft design that permits adjustment of shaft length and impeller depth—allowing you to place or remove mixing vessels without repositioning the mixer. The motor and drive unit are conveniently integrated into a compact aluminum and thermoplastic housing with easy-to-use controls and a clear LED display of rpm. Mixers feature infinitely adjustable stepless speed control. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in fluid viscosity.
Quiet, fan-cooled motors feature thermal overload shut-off, and an efficient heat-dissipating design to ensure a long service life. Motor is maintenance-free and rated for continuous duty. Safety circuits ensure automatic cut-off in an anti-stall or overload condition.
352,641.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
IKA 3990001 Digital Mixer, Dual-Range 6 to 400 / 30 to 2000 rpm, 115 VAC 5070440 Mfr # 3990001
COMPARE
2000 283 115
Call For Price
IKA 3990000 Digital Mixer, Dual-Range 6 to 400 / 30 to 2000 rpm, 230 VAC 5070445 Mfr # 0003990000
COMPARE
2000 283 230
₹352,641.00INR / Each