IKA EUROSTAR 20 High-Speed Digital Mixer
Mixing speeds up to 6000 rpm allow for rapid mixing
  • High-performance mixer for volumes up to 5.3 gallons (20 liters) and viscosities up to 10,000 cps
  • Precision cone seat ensures stirring shaft stability through full speed range
These mixers feature a motor and drive unit conveniently integrated into a compact aluminum and thermoplastic housing. Easy-to-use controls and a clear LED display of rpm enable you to precisely control and repeat your process parameters. Mixers feature infinitely adjustable stepless speed control. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in fluid viscosity.
Quiet, fan-cooled motors feature thermal overload shut-off, and an efficient heat-dissipating design to ensure a long service life. Motor is maintenance-free and rated for continuous duty. Safety circuits ensure automatic cut-off in an anti-stall or overload condition and also produce safe, smooth starts to prevent spraying.
232,811.00 - ₹854,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
IKA Eurostar 20 High-Speed Digital Mixer, 150 to 6000 rpm, 115 VAC 5070260 Mfr # 4028601
COMPARE
6000 28.3 115
₹854,000.00INR / Each
IKA Eurostar 20 High-Speed Digital Mixer, 150 to 6000 rpm, 230 VAC 5070265 Mfr # 0004028600
COMPARE
6000 28.3 230
₹232,811.00INR / Each