IKA® Digital Ceramic-Top Hot Plate Stirrers
Precise digital control with superior stirring strength
  • Exact temperature and speed setting via digital display
  • Extremely fast heating
Easy to clean and acid and alkalies resistant white ceramic top plate. Units feature three unique modes of operation guarantee optimal adjustment to your working method: 1) Mode A is suitable for work with varying parameters and is safety temperature adjustable 2) Mode B is suitable for series operation under uniform conditions 3) Mode C is suitable for unsupervised operation.
Microprocessor-controlled stirring hot plates feature an over-temperature safety circuit fixed at 1022°F (550°C).
58,384.00 - ₹236,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Power (VAC) Top Plate Length (cm)
Availability Pricing
IKA 3581001 Ceramic Stirring Hot Plate, 4" x 4", 115 VAC WW-04671-26 Mfr # 3581001
COMPARE
1500 115 10.16
₹185,000.00INR / Each
IKA 3581000 Ceramic Stirring Hot Plate, 4" x 4", 230 VAC WW-04671-28 Mfr # 0003581000
COMPARE
1500 230 10.16
₹58,384.00INR / Each
IKA C-Mag HS Digital Stirring Hot Plate, 7 x 7", 115 VAC WW-04671-30 Mfr # 3581201
COMPARE
1500 115 17.78
₹192,000.00INR / Each
IKA C-Mag HS Digital Stirring Hot Plate, 7 x 7", 230 VAC WW-04671-32 Mfr # 0003581200
COMPARE
1500 230 17.78
₹59,731.00INR / Each
IKA 3581401 Ceramic Stirring Hot Plate, 10" x 10", 115 VAC WW-04671-34 Mfr # 3581401
COMPARE
1500 115 25.4
₹236,000.00INR / Each
IKA 3581400 Ceramic Stirring Hot Plate, 10" x 10", 230 VAC WW-04671-36 Mfr # 3581400
COMPARE
1500 230 25.4
₹87,575.00INR / Each