IKA C5000 Automated Calorimeter and Accessories
Adiabatic, isoperibolic, and dynamic measurements - all in one system!
  • Fully automated system for easy GCV determination on solids or liquids
  • Integrated cooling system
  • Easy-to-use software
Used to measure heat released or absorbed during a process, bomb calorimeters calculate the Gross Calorifi c Value (GCV) of a solid or liquid. The IKA C 5000 is the only bomb calorimeter to feature three separate GCV determination methods in one instrument: adiabatic, isoperibolic, and dynamic.
Generally, measurements are performed using adiabatic or isoperibolic methods. Adiabatic methods minimize heat loss by keeping the surrounding temperature the same as the sample temperature, making it the most accurate for measuring calorific values of solids or liquids. Isoperibolic methods allow the reaction within the vessel to take place, while the water temperature in the surrounding vessel is held constant. Any temperature differential is compensated for once the reaction is complete. The dynamic mode is a shorter version of the adiabatic and/or isoperibol measuring principles. Results still conform to the required precision of the official standards, in accordance with DIN 51900, ASTM D240, ISO 1928, and BSI standards.
Fully Automated System
The IKA C5000 bomb calorimeter includes CalWin software, allowing the user to program, control, and analyze data. Automated water handling, oxygen filling, degassing, and the identification of decomposition vessels require no monitoring by the end-user. Connect calorimeter to scale, printer, PC, or LIMS software via RS-232 ports. Large internal memory holds up to 1000 separate test methods.
Wide Variety of Applications
Applications include fuels, foods, animal and plant products, and waste disposal. The IKA C5000 allows you to test just about any product from paraffin to pellets of cellulose.
370.00 - ₹593,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Measurement Reproducibility Measurement Types Measuring time
Availability Pricing
IKA 7114000 Automated Decomosition Vessel 5080570 Mfr # 7114000
COMPARE
₹593,000.00INR / Each
IKA 7215000 Halogen Resistant Decomposition Vessel 5080572 Mfr # 7215000
COMPARE
Call For Price
IKA 7207700 Calorimeter Aqua Pro Solution 5080574 Mfr # 7207700
COMPARE
₹5,500.00INR / Each
IKA 1483700 Cotton Thread 500 Pieces 5080576 Mfr # 1483700
COMPARE
₹5,500.00INR / Pkg of 1
IKA 7122800 Ignition Wire Pack (5/pk) 5080578 Mfr # 7122800
COMPARE
₹5,991.00INR / Pkg of 5
IKA 1749500 C5 Combustion Crucibles (24/pk) 5080580 Mfr # 1749500
COMPARE
₹21,252.00INR / Pkg of 24
IKA 3243000 Benzoic Acid Pellets (50/pk) 5080584 Mfr # 3243000
COMPARE
₹16,803.00INR / Pkg of 50
IKA 3041800 Calorimeter piston accessory 5080586 Mfr # 3041800
COMPARE
₹22,500.00INR / Each
IKA 1740300 O Ring, 2 X 1.6 For C 5000 5080590 Mfr # 1740300
COMPARE
₹500.00INR / Each
IKA 3019800 Colormeter contact spring 5080592 Mfr # 3019800
COMPARE
₹2,500.00INR / Each
IKA 1583500 O Ring, Viton, 4 X 1.5 mm For C 5000 5080594 Mfr # 1583500
COMPARE
₹530.00INR / Each
IKA 15836000 O Ring, Viton, 3.1x1.6 For C 5003 5080596 Mfr # 15836000
COMPARE
₹370.00INR / Each
Showing 1- 12 of 12