Idex® VHP (Very High Pressure) Reusable Chromatography Fittings
Fittings for very high pressure situations
  • Ideal for UHPLC applications Quick component replacement with minimal down time
The most demanding HPLC applications require fittings that will meet higher pressure requirements than standard chromatography fittings can accommodate. This patented line of VHP fittings are perfect for use in high-performance analytical systems. The VHP reusable fittings can be up to ten repeat assembly cycles without impacting pressure-holding ability when proper torque tightening specifications are followed. Increased component life can be achieved with a polymer front ferrule, which will not damage the tubing or receiving port.
14,800.00 - ₹160,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 1) (InchShort) Description
Availability Pricing
Idex VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 1/EA 0202272 Mfr # VHP-320
COMPARE
1/16 10-32 coned VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 1/EA
₹14,800.00INR / Each
Idex VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/2" Knurl; 1/EA 0202273 Mfr # VHP-3200
COMPARE
1/16 10-32 coned VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/2" Knurl; 1/EA
₹16,000.00INR / Each
Idex VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/2" Knurl; 10/PK 0202274 Mfr # VHP-3200X
COMPARE
1/16 10-32 coned VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/2" Knurl; 10/PK
₹160,000.00INR / Pkg of 10
Idex VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 10/PK 0202275 Mfr # VHP-320X
COMPARE
1/16 10-32 coned VHP Reusable Fitting, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 10/PK
₹144,000.00INR / Pkg of 10
Idex VHP Reusable Fitting, Long, SS/Pk, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 1/Ea 0202277 Mfr # VHP-325
COMPARE
1/16 10-32 coned VHP Reusable Fitting, Long, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 1/EA
₹14,800.00INR / Each
Idex VHP Reusable Fitting, Long, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 10/PK 0202278 Mfr # VHP-325X
COMPARE
1/16 10-32 coned VHP Reusable Fitting, Long, SS/PK, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 10/PK
₹151,000.00INR / Pkg of 10