Idex® VHP (Very High Pressure) Chromatography Stainless Steel Fittings
Designed to withstand extreme pressures
  • Ideal for UHPLC applications Two compression points provide twice the grip of standard ferrule
The most demanding HPLC applications require fittings that will meet higher pressure requirements than standard chromatography fittings can accommodate. This patented line of VHP fittings are perfect for use in high-performance analytical systems. The unique double compression ferrule design does not restrict the inner diameter of tubing and allows a less concentrated bite on the tubing.
3,800.00 - ₹34,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 1) (in) Description
Availability Pricing
Idex VHP Fitting, Male Nut/Ferrule, SS, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 1/EA WW-02022-66 Mfr # VHP-200
COMPARE
1/16 10-32 coned VHP Fitting, Male Nut/Ferrule, SS, 1/16\" OD Tubing, 10-32 Coned, 1/4\" Hex; 1/EA
₹3,800.00INR / Each
Idex VHP Fitting, Male Nut/Ferrule, SS, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 10/PK WW-02022-70 Mfr # VHP-200X
COMPARE
1/16 10-32 coned VHP Fitting, Male Nut/Ferrule, SS, 1/16\" OD Tubing, 10-32 Coned, 1/4\" Hex; 10/PK
₹34,700.00INR / Pkg of 10
Idex VHP Fitting, Male Nut/Ferrule, Long, SS, 1/16" OD Tubing, 10-32 Coned, 1/4" Hex; 1/EA WW-02022-71 Mfr # VHP-205
COMPARE
1/16 10-32 coned VHP Fitting, Male Nut/Ferrule, Long, SS, 1/16\" OD Tubing, 10-32 Coned, 1/4\" Hex; 1/EA
₹4,700.00INR / Each