Idex® Ultra High-Pressure MicroTight® Adapter Bodies
Create a high-pressure connection between 1/16" OD tubing and capillary tubing
  • Securely join together fluid pathway components
  • Manufactured from 316 stainless steel
  • Holds tight connection even through vibration
Port 1 is 10-32 coned for 1/16" OD tubing.
25,700.00 - ₹37,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Thread Size ISO Metric (F) (Port 2) (mm) Thread size UNF(F) (port 2) (InchShort)
Availability Pricing
Idex High-Pressure Adapter Body, SS, 1/16" to 1/32" OD Tubing, 10-32 to M4 0202417 Mfr # 1958-01
COMPARE
4
₹25,700.00INR / Each
Idex Ultra High-Pressure Adapter Body, SS, 1/16" to 1/32" OD Tubing, 10-32 to 6-40 0202510 Mfr # UH-631-01
COMPARE
6-40 Coned
₹37,400.00INR / Each