Idex® Super Flangeless™ Nuts, 5/16-24 Flat Bottom
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1,800.00 - ₹17,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Description
Availability Pricing
Idex Super Flangeless™ Nut, Standard Knurl, Black PEEK, 3/16" OD Tubing, 5/16-24 Flat Bottom; 1/EA 0202138 Mfr # P-137
COMPARE
Black 3/16 Super Flangeless™ Nut, Standard Knurl, Black PEEK, 3/16" OD Tubing, 5/16-24 Flat Bottom; 1/EA
₹1,800.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Black PEEK, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK 0202139 Mfr # P-137X
COMPARE
Black 3/16 Super Flangeless™ Nut, Standard Knurl, Black PEEK, 3/16" OD Tubing, 5/16-24 Flat Bottom; 10/PK
₹17,700.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/16" OD Tubing, 5/16-24 Flat Bottom; 1/EA 0202142 Mfr # P-141
COMPARE
Natural 1/16 Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/16" OD Tubing, 5/16-24 Flat Bottom; 25/PK
₹8,600.00INR / Each