Idex® Super Flangeless™ Nuts, 1/4-28 Flat Bottom for 1/8" OD Tubing
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
490.00 - ₹10,700.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0201380 Mfr # P-331X
COMPARE
Natural PEEK Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹10,700.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0201381 Mfr # P-336X
COMPARE
Natural PEEK Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹10,700.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Natural PPS, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0201386 Mfr # P-381
COMPARE
Natural PPS Super Flangeless™ Nut, Standard Knurl, Natural PPS, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,000.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202180 Mfr # P-331
COMPARE
Natural PEEK Super Flangeless™ Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Black PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202182 Mfr # P-331BLK
COMPARE
Black PEEK Super Flangeless™ Nut, Standard Knurl, Black PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Green PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202183 Mfr # P-331G
COMPARE
Green PEEK Super Flangeless™ Nut, Standard Knurl, Green PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,300.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Orange PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202184 Mfr # P-331OR
COMPARE
Orange PEEK Super Flangeless™ Nut, Standard Knurl, Orange PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 25/PK
₹2,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Yellow PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202185 Mfr # P-331Y
COMPARE
Yellow PEEK Super Flangeless™ Nut, Standard Knurl, Yellow PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 25/PK
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Black POM, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202186 Mfr # P-332
COMPARE
Black POM Super Flangeless™ Nut, Standard Knurl, Black POM, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹490.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Blue POM, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202187 Mfr # P-332B
COMPARE
Blue POM Super Flangeless™ Nut, Standard Knurl, Blue POM, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹490.00INR / Each
Idex Super Flangeless™ Nut, Standard Knurl, Black POM, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202188 Mfr # P-332X
COMPARE
Black POM Super Flangeless™ Nut, Standard Knurl, Black POM, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹4,700.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202189 Mfr # P-336
COMPARE
Natural PEEK Super Flangeless™ Nut, Headless, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹1,200.00INR / Each
Idex Super Flangeless™ Nut, Headless, Natural PPS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202208 Mfr # P-387
COMPARE
Natural PPS Super Flangeless™ Nut, Headless, Natural PPS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
₹1,000.00INR / Each
Idex Super Flangeless™ Nut, Headless, Natural PPS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202209 Mfr # P-387X
COMPARE
Natural PPS Super Flangeless™ Nut, Headless, Natural PPS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
₹9,500.00INR / Pkg of 10
Idex Super Flangeless™ Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202210 Mfr # P-400
COMPARE
Natural PFA Super Flangeless™ Nut, Standard Knurl, Natural PFA, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
₹1,400.00INR / Each
Showing 1- 15 of 15