Idex® Super Flangeless™ Fitting Assemblies, M6 Flat Bottom
Flat-bottom fitting assemblies withstand up to 5000 psi (345 bar)!
  • Eliminates tubing twist that can loosen connections
  • Resistant to loosening through vibration
Super Flangeless™ fitting assemblies are an excellent choice for tubing assemblies and will stay tight even through vibration. One-piece PEEK fittings are reusable that require no swaging. Two-piece fittings come with ferrules with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1,600.00 - ₹56,600.00INR / Pkg of 25
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/16" OD Tubing, M6 Flat Bottom; 25/PK 0202149 Mfr # P-229
COMPARE
1/16 PEEK Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/16" OD Tubing, M6 Flat Bottom
₹12,300.00INR / Pkg of 25
Idex Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/8" OD Tubing, M6 Flat Bottom; 1/EA 0202178 Mfr # P-329
COMPARE
1/8 PEEK Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/8" OD Tubing, M6 Flat Bottom
₹15,400.00INR / Each
Idex Super Flangeless™ Fitting, Black PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 1/EA 0202198 Mfr # P-357
COMPARE
5/64 PEEK/SS Super Flangeless™ Fitting, Black PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 1/EA
₹4,700.00INR / Each
Idex Super Flangeless™ Fitting, Short, Black PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 1/EA 0202199 Mfr # P-357-1
COMPARE
5/64 PEEK/SS Super Flangeless™ Fitting, Short, Black PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom
₹1,600.00INR / Each
Idex Super Flangeless™ Fitting, Black PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 10/PK 0202203 Mfr # P-357X
COMPARE
5/64 PEEK/SS Super Flangeless™ Fitting, Black PEEK/SS Ring, 2 mm OD Tubing, M6 Flat Bottom; 10/PK
₹56,600.00INR / Pkg of 10