Idex® Super Flangeless™ Fitting Assemblies, 1/4-28 and 5/16-24 Flat Bottom
Flat-bottom fitting assemblies withstand high pressures
  • Eliminates tubing twist that can loosen connections
  • Resistant to loosening through vibration
Super Flangeless™ fitting assemblies are an excellent choice for tubing assemblies and will stay tight even through vibration. One-piece PEEK fittings are reusable that require no swaging. Two-piece fittings come with ferrules with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
3,000.00 - ₹39,500.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 1) (InchShort) Description
Availability Pricing
Idex Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202156 Mfr # P-249
COMPARE
1/16 1/4-28 flat bottom Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹3,700.00INR / Each
Idex Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202157 Mfr # P-249X
COMPARE
1/16 1/4-28 flat bottom Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹39,500.00INR / Pkg of 10
Idex Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202191 Mfr # P-349
COMPARE
1/8 1/4-28 flat bottom Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹3,000.00INR / Each
Idex Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK 0202192 Mfr # P-349X
COMPARE
1/8 1/4-28 flat bottom Super Flangeless™ Fitting, One Piece, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
₹26,700.00INR / Pkg of 10
Idex Super Flangeless™ Fitting, Black PEEK/ETFE/SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 1/EA 0202214 Mfr # XP-137
COMPARE
3/16 5/16-24 flat bottom Super Flangeless™ Fitting, Black PEEK/ETFE/SS, 3/16" OD Tubing, 5/16-24 Flat Bottom; 25/PK
₹3,300.00INR / Each
Idex Super Flangeless™ Fitting, Natural PEEK/ETFE/SS, 1/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA 0202216 Mfr # XP-141
COMPARE
1/16 5/16-24 flat bottom Super Flangeless™ Fitting, Natural PEEK/ETFE/SS, 1/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA
₹9,000.00INR / Each