IDEX® Stainless Steel Tubing, 1/8" OD
Precut stainless steel tubing ready for all chromatography testing requirements
  • 316 stainless steel tubing, polished and passivated
  • Color coded for easy identification
  • Suited for most analytical applications
IDEX 316 stainless steel tubing is precut and thoroughly passivated (except lengths 25 feet and beyond), making it the recommended choice for most chromatographic analytical applications. Tubing is machine cut, polished. and each end is deburred inside and out. Then it is submerged in an acid passivation bath and flushed clean. Each tube is color coded for easy identification of inner diameter.
9,600.00 - ₹156,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Pressure (PSI) Tubing ID (in)
Availability Pricing
Idex U-800 Chromatography Tubing, 316 SS, 1/8" OD x 0.080" ID x 3.2 ft L, Natural; 1/EA 0201174 Mfr # U-800
COMPARE
3.2 7600 5/64
₹32,600.00INR / Each
Idex U-803 Chromatography Tubing, 316 SS, 1/8" OD x 0.080" ID x 9.8 ft L, Natural; 1/EA 0201175 Mfr # U-803
COMPARE
9.8 7600 5/64
₹93,600.00INR / Each
Idex U-805 Chromatography Tubing, 316 SS, 1/8" OD x 0.080" ID x 16 ft L, Natural; 1/EA 0201176 Mfr # U-805
COMPARE
16 7600 5/64
₹156,000.00INR / Each
Idex U-825 Chromatography Tubing, 316 SS, 1/8" OD x 0.080" ID x 10" L, Natural; 1/EA 0201177 Mfr # U-825
COMPARE
0.833 7600 5/64
₹9,600.00INR / Each