IDEX® Stainless Steel Tubing, 1/32" OD
Precut stainless steel tubing ready for all chromatography testing requirements
  • 316 stainless steel tubing, polished and passivated
  • Color coded for easy identification
  • Suited for most analytical applications
IDEX 316 stainless steel tubing is precut and thoroughly passivated (except lengths 25 feet and beyond), making it the recommended choice for most chromatographic analytical applications. Tubing is machine cut, polished. and each end is deburred inside and out. Then it is submerged in an acid passivation bath and flushed clean. Each tube is color coded for easy identification of inner diameter.
3,400.00 - ₹13,500.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Pressure (PSI) Tubing ID (in)
Availability Pricing
Idex U-1113 Chromatography Tubing, 316 SS, 1/32" OD x 0.003" ID x 0.98" L, Red; 1/EA 0201091 Mfr # U-1113
COMPARE
0.083 19300 1/300
₹4,900.00INR / Each
Idex Chromatography Tubing, 316 Stainless Steel, 1/32" OD x 0.004" ID x 2" L, Red 0201092 Mfr # U-1114
COMPARE
0.167 19300 1/250
₹6,100.00INR / Each
Idex Chromatography Tubing, 316 Stainless Steel, 1/32" OD x 0.004" ID x 8" L, Red 0201094 Mfr # U-1116
COMPARE
0.666 19300 1/250
₹9,900.00INR / Each
Idex U-1117 Chromatography Tubing, 316 SS, 1/32" OD x 0.004" ID x 12" L, Red; 1/EA 0201095 Mfr # U-1117
COMPARE
1 19300 1/250
₹11,400.00INR / Each
Idex U-1120 Chromatography Tubing, 316 SS, 1/32" OD x 0.006" ID x 2" L, Yellow; 1/EA 0201097 Mfr # U-1120
COMPARE
0.167 19300 1/175
₹4,700.00INR / Each
Idex U-1122 Chromatography Tubing, 316 SS, 1/32" OD x 0.006" ID x 8" L, Yellow; 1/EA 0201098 Mfr # U-1122
COMPARE
0.666 19300 1/175
₹9,300.00INR / Each
Idex U-1123 Chromatography Tubing, 316 SS, 1/32" OD x 0.006" ID x 11.8" L, Yellow; 1/EA 0201099 Mfr # U-1123
COMPARE
0.983 19300 1/175
₹8,800.00INR / Each
Idex U-1125 Chromatography Tubing, 316 SS, 1/32" OD x 0.008" ID x 2" L, Clear; 1/EA 0201102 Mfr # U-1125
COMPARE
0.167 17800 1/125
₹4,500.00INR / Each
Idex U-1126 Chromatography Tubing, 316 SS, 1/32" OD x 0.008" ID x 4" L, Clear; 1/EA 0201103 Mfr # U-1126
COMPARE
0.333 17800 1/125
₹4,200.00INR / Each
Idex U-1130 Chromatography Tubing, 316 SS, 1/32" OD x 0.010" ID x 2" L, Blue; 1/EA 0201106 Mfr # U-1130
COMPARE
0.167 16200 1/100
₹3,900.00INR / Each
Idex U-1140 Chromatography Tubing, 316 SS, 1/32" OD x 0.015" ID x 2" L, Green; 1/EA 0201113 Mfr # U-1140
COMPARE
0.167 12300 1/64
₹4,700.00INR / Each
Idex U-1141 Chromatography Tubing, 316 SS, 1/32" OD x 0.015" ID x 4" L, Green; 1/EA 0201114 Mfr # U-1141
COMPARE
0.333 12300 1/64
₹6,700.00INR / Each
Idex U-1142 Chromatography Tubing, 316 SS, 1/32" OD x 0.015" ID x 8" L, Green; 1/EA 0201115 Mfr # U-1142
COMPARE
0.666 12300 1/64
₹10,000.00INR / Each
Idex U-1143 Chromatography Tubing, 316 SS, 1/32" OD x 0.015" ID x 12" L, Green; 1/EA 0201116 Mfr # U-1143
COMPARE
1 12300 1/64
₹13,500.00INR / Each
Idex U-1145 Chromatography Tubing, 316 SS, 1/32" OD x 0.018" ID x 2" L, Black; 1/EA 0201117 Mfr # U-1145
COMPARE
0.167 10000 1/64
₹3,400.00INR / Each
Idex U-1146 Chromatography Tubing, 316 SS, 1/32" OD x 0.018" ID x 4" L, Black; 1/EA 0201118 Mfr # U-1146
COMPARE
0.333 10000 1/64
₹6,700.00INR / Each
Showing 1- 16 of 16