Idex® Stainless Steel Sample Loops for Injection Valve Models 7125 and 7010
Square-cut, burr-free ends provide a firm, flush connection to valve ports
  • 316 stainless steel construction
  • High pressure resistance, low permeability
A sample loop is a defined length of tubing and fittings configured to match the angle of the injection valve ports. A sample is loaded and held in the loop until injection is triggered, either automatically or manually. Stainless steel loops are supplied with unswaged fittings, so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal.
16,600.00 - ₹72,600.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (µL) Tubing Inside Diameter (ID) (mm) Tubing Outside Diameter (OD) (in)
Availability Pricing
Idex Sample Loop for 7125, 7010, 7725, and 7725i Valves, SS, 10 mL, 1/8" OD x 0.080" ID; 1/EA 0201632 Mfr # 1876
COMPARE
10000 2 0.125
₹72,600.00INR / Each
Idex Sample Loop for 7125/7010 Valves, SS, 5 µL, 1/16" OD x 0.007" ID; 1/EA 0201633 Mfr # 7020
COMPARE
5 0.18 0.0625
₹19,100.00INR / Each
Idex Sample Loop for 7125/7010 Valves, Stainless Steel, 10 µL, 1/16" OD x 0.012" ID 0201634 Mfr # 7021
COMPARE
10 0.3 0.0625
₹18,000.00INR / Each
Idex Sample Loop for 7125/7010 Valves, Stainless Steel, 20 µL, 1/16" OD x 0.020" ID 0201635 Mfr # 7022
COMPARE
20 0.51 0.0625
₹16,600.00INR / Each
Idex Sample Loop for 7125/7010 Valves, Stainless Steel, 50 µL, 1/16" OD x 0.020" ID 0201636 Mfr # 7023
COMPARE
50 0.51 0.0625
₹19,200.00INR / Each
Idex Sample Loop for 7125/7010 Valves, SS, 100 µL, 1/16" OD x 0.020" ID; 1/EA 0201637 Mfr # 7024
COMPARE
100 0.51 0.0625
₹18,600.00INR / Each
Idex Sample Loop for 7125/7010 Valves, SS, 500 µL, 1/16" OD x 0.030" ID; 1/EA 0201639 Mfr # 7026
COMPARE
500 0.76 0.0625
₹21,800.00INR / Each
Idex Sample Loop for 7125/7010 Valves, SS, 1 mL, 1/16" OD x 0.030" ID; 1/EA 0201640 Mfr # 7027
COMPARE
1000 0.76 0.0625
₹25,100.00INR / Each
Idex Sample Loop for 7125/7010 Valves, SS, 5 mL, 1/16" OD x 0.040" ID; 1/EA 0201642 Mfr # 7029
COMPARE
5000 1 0.0625
₹64,300.00INR / Each
Showing 1- 9 of 9