Idex® Semi-Prep Solvent Filtration Frits
Extend filtration life and consistency with replaceable frits
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
Easily replaceable frits save you time and money lost to frit-cleaning labor, plus eliminate the problem of embedded debris re-entering your instrument. These frits are available in the most common HPLC filtration ratings. Each includes a PCTFE or PEEK polymer sealing ring, which are slightly thicker than the frit disc for extra sealing and chemical resistance. Many also have the most common outer diameters — 0.250" and 0.254" — making them ideal for use in inline or precolumn filters.
2,000.00 - ₹8,700.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
Idex Semi-Prep Frit, SS, 0.5 µm; 0.062" OD, 0.20" Black PCTFE Ring; 1/EA 0200926 Mfr # A-418
COMPARE
1.57 Stainless Steel 2
₹8,700.00INR / Each
Idex Replacement Frit, SS, 2 µm, 0.750" OD, 0.880" OD ETFE Ring, 141.9 µL; 1/EA 0200927 Mfr # A-332
COMPARE
19 Stainless Steel 2
₹5,900.00INR / Each
Idex Semi-Prep Frit, SS, 20 µm; 3/16" OD, 0.254" PTFE Ring; 1/EA 0201002 Mfr # A-224
COMPARE
4.8 Stainless Steel 20
₹2,100.00INR / Each
Idex Semi-Prep Frit, SS, 10 µm; 0.094" OD, 0.25" PCTFE Ring; 1/EA 0201056 Mfr # A-106
COMPARE
2.39 Stainless Steel 10
₹3,200.00INR / Each
Idex Semi-Prep Frit, SS, 10 µm; 0.189" OD, 0.254" PCTFE Ring; 1/EA 0201057 Mfr # A-107
COMPARE
4.8 Stainless Steel 10
₹3,200.00INR / Each
Idex Semi-Prep Frit, SS, 20 µm; 0.125" OD, 0.25" PCTFE Ring; 1/EA 0201058 Mfr # A-120
COMPARE
3.18 Stainless Steel 20
₹2,400.00INR / Each
Idex Semi-Prep Frit, SS, 20 µm; 0.188" OD, 0.254" PCTFE Ring; /EA 0201059 Mfr # A-122
COMPARE
4.8 Stainless Steel 20
₹2,000.00INR / Each
Idex Semi-Prep Frit, SS, 2 µm; 0.062" OD, 0.20" Green PCTFE Ring; 1/EA 0201062 Mfr # A-417
COMPARE
1.57 Stainless Steel 0.5
₹4,500.00INR / Each
Idex Semi-Prep Frit, PEEK, 10 µm, 0.125" OD, 0.25" PCTFE Ring, 4.2 µL 0201072 Mfr # A-720
COMPARE
3.18 PEEK 10
₹3,500.00INR / Each
Idex Semi-Prep Frit, PEEK, 10 µm, 0.197" OD, 0.25" PCTFE Ring, 9.9 µL 0201073 Mfr # A-722
COMPARE
5 PEEK 10
₹3,500.00INR / Each
Idex Semi-Prep Frit, PEEK, 5 µm; 0.293" OD, 0.375" PCTFE Ring; 1/EA 0201081 Mfr # OC-815
COMPARE
7.44 PEEK 5
₹5,900.00INR / Each
Idex Semi-Prep Frit, SS, 20 µm; 0.750" OD, 0.880" OD ETFE Ring, 152.0 µL; 1/EA 4271159 Mfr # A-337
COMPARE
19 Stainless Steel 20
₹6,800.00INR / Each
Idex Biocompatible Frit, PEEK, 2 µm; 0.460" OD, 0.56" Green PCTFE Ring, 46.4 µL; 1/EA 4271165 Mfr # OC-802
COMPARE
11.7 PEEK 2
₹7,000.00INR / Each
Idex Biocompatible Frit, PEEK, 10 µm; 0.460" OD, 0.56" PCTFE Ring, 57.2 µL; 1/EA 4271166 Mfr # OC-803
COMPARE
11.7 PEEK 10
₹7,100.00INR / Each
Idex Biocompatible Frit, PEEK, 5 µm, 0.460" OD, 0.56" OD PCTFE Ring, 41.1 µL; 1/EA 4271167 Mfr # OC-805
COMPARE
11.7 PEEK 5
₹7,000.00INR / Each
Showing 1- 15 of 15