Idex® Semi-Prep Solvent Filters
Protect larger columns and injections from harmful particulates
  • Ideal for higher-flow systems
  • Interchangeable frits make protection more economical
  • Compatible with SFC and HPLC
  • Available to fit a variety of tubing sizes
Designed for high-flow applications, these filters are ideal for prep, semi-prep, and analytical processes. When the interchangeable frits become clogged, they can be easily replaced by unscrewing the assembly and replacing the frit.
4,500.00 - ₹92,600.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (µm) Membrane Material Description
Availability Pricing
Idex Iso-Prep Filter Holder, 21.2 mm, 0.5 µm SS Frit, 10-32 coned port; 1/EA WW-42711-52 Mfr # 1020-05
COMPARE
0.5 Stainless Steel Iso-Prep Filter Holder, 21.2 mm, SS 0.5 µm Frit, 10-32 coned port; 203 µL
₹78,600.00INR / Each
Idex Iso-Prep Filter Holder, 21.2 mm, 2 µm SS Frit, 10-32 coned port; 1/EA WW-42711-53 Mfr # 1020-20
COMPARE
2 Stainless Steel Iso-Prep Filter Holder, 21.2 mm, 2 µm SS Frit, 10-32 coned port; 1/EA
₹92,600.00INR / Each
Idex Iso-Prep Filter, SS, 0.5 µm, 21.2 mm; 1/EA WW-42711-54 Mfr # 7031-05
COMPARE
0.5 Stainless Steel Iso-Prep Filter, SS, 0.5 µm, 21.2 mm; 1/EA
₹14,500.00INR / Each
Idex Iso-Prep Filter, SS, 2 µm, 21.2 mm; 1/EA WW-42711-55 Mfr # 7031-20
COMPARE
2 Stainless steel Iso-Prep Filter, SS, 2 µm, 21.2 mm; 1/EA
₹4,500.00INR / Each
Idex Semi-Prep Filter Assembly, SS/PEEK, 10 µm, 10-32 Coned; 1/EA WW-42711-56 Mfr # A-330
COMPARE
10 Stainless Steel Semi-Prep Filter Assembly, SS/PEEK, 10 µm, 10-32 Coned; 1/EA
₹25,300.00INR / Each
Idex Semi-Prep Filter Assembly, SS, 10 µm, 5/16-24 Flat Bottom; 1/EA WW-42711-60 Mfr # A-360
COMPARE
10 Stainless Steel Semi-Prep Filter Assembly, SS, 10 µm, 5/16-24 Flat Bottom; 1/EA
₹45,900.00INR / Each
Idex Biocompatible Semi-Prep Filter Assembly, SS & PEEK, 2 µm, 10-32 Coned; 1/EA WW-42711-61 Mfr # A-410
COMPARE
2 PEEK Biocompatible Semi-Prep Filter Assembly, SS & PEEK, 2 µm, 10-32 Coned; 1/EA
₹25,700.00INR / Each
Idex Biocompatible Semi-Prep Filter Assembly, SS, 10 µm PEEK/PCTFE Frit, 10-32 Coned; 1/EA WW-42711-62 Mfr # A-411
COMPARE
10 PEEK Biocompatible Semi-Prep Filter Assembly, SS, 10 µm PEEK/PCTFE Frit, 10-32 Coned; 1/EA
₹25,700.00INR / Each
Idex Biocompatible Semi-Prep Filter Assembly, SS, 5 µm PEEK Frit, 5/16-24 Flat Bottom; 1/EA WW-42711-63 Mfr # A-510
COMPARE
5 PEEK Biocompatible Semi-Prep Filter Assembly, SS, 5 µm PEEK Frit, 5/16-24 Flat Bottom; 1/EA
₹26,500.00INR / Each
Showing 1- 9 of 9