Idex® RheFlex® Fittings
Simplify by getting an all-in-one fitting for your fluidic needs
  • PEEK construction for an inert, biocompatible connection
  • Color-coded ChromTRAC knobs for easy identification
A. One-Piece RheFlex M4 Fitting
Eliminates the need for separate nuts and ferrules
The combination of a one-piece nut and ferrule makes connections easier and faster. The fitting features a unique seal-tight grip. When used with PEEK tubing sleeve, the fitting can hold up to 5000 psi (345 bar). They are compatible with ChromTRAC knobs.
B. Two-Piece RheFlex Finger-Tight Fittings
No need for tools to tighten these fittings
Included ChromTRAC knobs allow for quick identification. The 10-32 coned bottom fittings are available in standard, short and extra-long lengths. Each fitting comes with a PEEK ferrule. The fittings are rated for use up to 7000 psi (483 bar).
8,200.00 - ₹30,100.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Fitting Length (mm)
Availability Pricing
Idex RheFlex® Replacement Ferrule, 1/16" OD Tubing, PEEK; 1/EA 0203215 Mfr # 6000-051
COMPARE
PEEK
₹8,200.00INR / Each
Idex RheFlex® Two-Piece Fitting, 1/16" OD Tubing, 10-32 Coned, PEEK; 1/EA 0203216 Mfr # 6000-054
COMPARE
PEEK
₹12,000.00INR / Each
Idex RheFlex® Fitting, Short, 1/16" OD Tubing, 10-32 Coned, PEEK; 1/EA 0203217 Mfr # 6000-055
COMPARE
PEEK
₹12,500.00INR / Each
Idex RheFlex® Two-Piece Finger-Tight Fitting, Extra Long, 1/16" OD Tubing, 10-32 Coned, Black, PEEK; 1/EA 0203218 Mfr # 6000-066
COMPARE
Black PEEK
₹9,700.00INR / Each
Idex RheFlex® Two-Piece Finger-Tight Fitting, 1/16" OD Tubing, 10-32 Coned, Yellow, PEEK; 10/PK 0203226 Mfr # 6000-254YL
COMPARE
Yellow PEEK
₹30,100.00INR / Pkg of 10