Idex® Quick Connect Female Luer-to-MicroTight Adapter
Convenience of quick connect and disconnect of a luer connection
  • Easily connect a syringe to 360 µm OD tubing
Directly connect 360 µm OD tubing without tubing sleeves to a luer-tip syringe or other products with a male luer connection. Fitting assembly comes with MicroTight® fittings.
20,100.00 - ₹20,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type-Port 1 Material Description
Availability Pricing
Idex Threaded Luer Adapter, Natural PEEK/PK, 0.006" Bore, Female Luer x MicroTight® Assembly; 1/EA 0201416 Mfr # P-662
COMPARE
Female luer PEEK/PK Threaded Luer Adapter, Natural PEEK/PK, 0.006" Bore, Female Luer x MicroTight® Assembly; 1/EA
₹20,300.00INR / Each
Idex Threaded Luer Adapter, Natural PEEK, 0.006" Bore, Female Luer x MicroTight® Body; 1/EA 0202361 Mfr # P-662-01
COMPARE
Female luer PEEK Threaded Luer Adapter, Natural PEEK, 0.006" Bore, Female Luer x MicroTight® Body; 1/EA
₹20,100.00INR / Each