Idex® PEEK Sample Loops for Injection Valve Models 9725, 9725i, 9010, PR/EV750-100, and PR/EV753-100
Biocompatible and inert to almost all organic solvents
  • Excellent alternative to stainless steel loops
  • Easy to install or replace
Biocompatible PEEK polymer sample loops are inert to almost all organic solvents, making them a versatile alternative to stainless steel. The loop ends are cut clean and straight for easy valve installation.
7,900.00 - ₹46,400.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (µL) Tubing Inside Diameter (ID) (mm) Tubing Outside Diameter (OD) (in)
Availability Pricing
Idex Sample Loop for 9725/9725i Valves, PEEK, 20 µL, 1/16" OD x 0.010" ID; 1/EA WW-02013-20 Mfr # 9055-022
COMPARE
20 0.25 0.0625
₹9,500.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 100 µL, 1/16" OD x 0.020" ID; 1/EA WW-02013-21 Mfr # 9055-024
COMPARE
100 0.51 0.0625
₹9,500.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 5 µL, 1/16" OD x 0.007" ID; 1/EA WW-02016-77 Mfr # 9055-020
COMPARE
5 1.8 0.0625
₹7,900.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 10 µL, 1/16" OD x 0.010" ID; 1/EA WW-02016-78 Mfr # 9055-021
COMPARE
10 0.25 0.0625
₹9,700.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 50 µL, 1/16" OD x 0.020" ID; 1/EA WW-02016-79 Mfr # 9055-023
COMPARE
50 0.51 0.0625
₹9,500.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 200 µL, 1/16" OD x 0.020" ID; 1/EA WW-02016-80 Mfr # 9055-025
COMPARE
200 0.51 0.0625
₹13,100.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 500 µL, 1/16" OD x 0.030" ID; 1/EA WW-02016-81 Mfr # 9055-026
COMPARE
500 0.76 0.0625
₹15,800.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 1 mL, 1/16" OD x 0.030" ID; 1/EA WW-02016-82 Mfr # 9055-027
COMPARE
1000 0.76 0.0625
₹17,800.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 2 mL, 1/16" OD x 0.030" ID; 1/EA WW-02016-83 Mfr # 9055-028
COMPARE
2000 0.76 0.0625
₹25,400.00INR / Each
Idex Sample Loop for 9725/9725i Valves, PEEK, 5 mL, 1/16" OD x 0.030" ID; 1/EA WW-02016-84 Mfr # 9055-029
COMPARE
5000 0.76 0.0625
₹46,400.00INR / Each
Showing 1- 10 of 10