Idex® Nonmetallic Inline Chromatography Check Valves
Metal-free, biocompatible materials are ideal for biological samples and corrosive fluids
  • Prevent back-flow to protect your system
  • Excellent chemical resistance
Featuring low swept volume, these nonmetallic check valves are perfect for applications where low path volume is critical. Multiple connection configuration offers easy adaptation to different applications.
38,900.00 - ₹80,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Inlet Port Orifice (in) Outlet Port
Availability Pricing
Idex Nonmetallic Inline Check Valve, Inlet, 1 psi, 0.02" ID, 1/4-28 (F) to 1/4-28 (M) Flat Bottom; 1/EA 0201336 Mfr # CV-3320
COMPARE
1/4-28 (F) flat bottom 0.02 1/4-28 (M) flat bottom
₹40,400.00INR / Each
Idex Nonmetallic Inline Check Valve, Outlet, 1 psi, 0.02" ID, 1/4-28 (M) to 1/4-28 (F) Flat Bottom; 1/EA 0201337 Mfr # CV-3321
COMPARE
1/4-28 (M) flat bottom 0.02 1/4-28 (F) flat bottom
₹40,400.00INR / Each
Idex Nonmetallic Inline Check Valve, Inlet, 1 psi, 0.06" ID, 1/4-28 (F) to 1/4-28 (M) Flat Bottom; 1/EA 0201338 Mfr # CV-3324
COMPARE
1/4-28 (F) flat bottom 0.06 1/4-28 (M) flat bottom
₹39,000.00INR / Each
Idex Nonmetallic Inline Check Valve, Outlet, 1 psi, 0.06" ID, 1/4-28 (M) to 1/4-28 (F) Flat Bottom; 1/EA 0201339 Mfr # CV-3325
COMPARE
1/4-28 (M) flat bottom 0.06 1/4-28 (F) flat bottom
₹38,900.00INR / Each
Idex Nonmetallic Inline Check Valve, Outlet, 1 psi, 0.02" ID, 10-32 (M) Coned to 1/4-28 (F) Flat Bottom; 1/EA 0201340 Mfr # CV-3336
COMPARE
10-32 (M) coned bottom 0.02 1/4-28 (F) flat bottom
₹80,000.00INR / Each
Idex Nonmetallic Inline Check Valve, Inlet, 1 psi, 0.04" ID, 1/4-28 (F) to 1/4-28 (M) Flat Bottom; 1/EA 0201591 Mfr # CV-3322
COMPARE
1/4-28 (F) flat bottom 0.04 1/4-28 (M) flat bottom
₹45,800.00INR / Each
Idex Nonmetallic Inline Check Valve, Outlet, 1 psi, 0.04" ID, 1/4-28 (M) to 1/4-28 (F) Flat Bottom; 1/EA 0201592 Mfr # CV-3323
COMPARE
1/4-28 (M) flat bottom 0.04 1/4-28 (F) flat bottom
₹43,700.00INR / Each
Idex Nonmetallic Inline Check Valve, 1 psi, 0.02" ID, 1/4-28 (F) to 1/4-28 (F) Flat Bottom; 1/EA 0201593 Mfr # CV-3330
COMPARE
1/4-28 (F) flat bottom 0.02 1/4-28 (F) flat bottom
₹39,900.00INR / Each
Idex Nonmetallic Inline Check Valve, Inlet, 1 psi, 0.02" ID, 1/4-28 (F) Flat Bottom to 10-32 (M) Coned; 1/EA 0201594 Mfr # CV-3335
COMPARE
1/4-28 (F) flat bottom 0.02 10-32 (M) coned bottom
₹44,400.00INR / Each
Idex Nonmetallic Inline Check Valve, 1 psi, 0.02" ID, 10-32 (F) to 10-32 (F) Coned; 1/EA 0201595 Mfr # CV-3340
COMPARE
10-32 (F) coned bottom 0.02 10-32 (F) coned bottom
₹42,000.00INR / Each
Idex Nonmetallic Inline Check Valve, Micro-Volume, 8 psi, 0.01" ID, 10-32 (F) to 10-32 (F) Coned; 1/EA 0201597 Mfr # CV-3500
COMPARE
10-32 (F) coned bottom 0.02 10-32 (F) coned bottom
₹68,800.00INR / Each
Showing 1- 11 of 11