IDEX® NanoTight™ FEP Capillary Tubing Sleeves
Ensure good port connection with capillary tubing
  • Maintain concentric connection with minimal dead volume
  • Color coded for easy identification
Sleeves are not permanently attached to a ferrule so they can easily adapt to varying tubing pocket depths minimizing dead volume. NanoTight™ FEP Tubing Sleeves are designed primarily for use with NanoTight fittings, but also work effectively with Super Flangeless™ fittings. Sleeves have a maximum recommended continuous operating temperature of 122°F (50°C) and withstand pressures up to 4000 psi (276 bar).
500.00 - ₹5,000.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Color Compatible With
Availability Pricing
Idex F-237 NanoTight™ Tubing Sleeve, Red FEP, 0.005" ID x 1/16" OD; 1/EA WW-02017-96 Mfr # F-237
COMPARE
Red 70 to 110 µm OD tubing
₹510.00INR / Each
Idex F-238 NanoTight™ Tubing Sleeve, Yellow FEP, 0.007" ID x 1/16" OD; 1/EA WW-02017-97 Mfr # F-238
COMPARE
Yellow 125 to 165 µm OD tubing
₹500.00INR / Each
Idex F-239 NanoTight™ Tubing Sleeve, Natural FEP, 0.0085" ID x 1/16" OD; 1/EA WW-02017-98 Mfr # F-239
COMPARE
Natural 160 to 200 µm OD tubing
₹500.00INR / Each
Idex F-240 NanoTight™ Tubing Sleeve, Blue FEP, 0.011" ID x 1/16" OD; 1/EA WW-02017-99 Mfr # F-240
COMPARE
Blue 225 to 265 µm OD tubing
₹510.00INR / Each
Idex F-241 NanoTight™ Tubing Sleeve, Orange FEP, 0.013" ID x 1/16" OD; 1/EA WW-02018-00 Mfr # F-241
COMPARE
Orange 275 to 315 µm OD tubing
₹500.00INR / Each
Idex F-241N NanoTight™ Tubing Sleeve, Natural FEP, 0.013" ID x 1/16" OD; 1/EA WW-02018-01 Mfr # F-241N
COMPARE
Natural 275 to 315 µm OD tubing
₹2,900.00INR / Each
Idex F-242 NanoTight™ Tubing Sleeve, Green FEP, 0.0155" ID x 1/16" OD; 1/EA WW-02018-02 Mfr # F-242
COMPARE
Green 340 to 380 µm OD tubing
₹500.00INR / Each
Idex F-242X NanoTight™ Tubing Sleeve, Green FEP, 0.0155" ID x 1/16" OD; 10/PK WW-02018-03 Mfr # F-242X
COMPARE
Green 340 to 380 µm OD tubing
₹4,900.00INR / Pkg of 10
Idex F-243 NanoTight™ Tubing Sleeve, Black FEP, 0.018" ID x 1/16" OD; 1/EA WW-02018-04 Mfr # F-243
COMPARE
Black 400 to 440 µm OD tubing
₹500.00INR / Each
Idex F-244 NanoTight™ Tubing Sleeve, Natural FEP, 0.021" ID x 1/16" OD; 1/EA WW-02018-05 Mfr # F-244
COMPARE
Natural 480 to 520 µm OD tubing
₹510.00INR / Each
Idex F-245 NanoTight™ Tubing Sleeve, Red FEP, 0.024" ID x 1/16" OD; 1/EA WW-02018-06 Mfr # F-245
COMPARE
Red 555 to 595 µm OD tubing
₹510.00INR / Each
Idex F-246 NanoTight™ Tubing Sleeve, Yellow FEP, 0.027" ID x 1/16" OD; 1/EA WW-02018-07 Mfr # F-246
COMPARE
Yellow 630 to 670 µm OD tubing
₹530.00INR / Each
Idex F-247 NanoTight™ Tubing Sleeve, Green FEP, 0.033" ID x 1/16" OD; 1/EA WW-02018-08 Mfr # F-247
COMPARE
Green 785 to 825 µm OD tubing
₹500.00INR / Each
Idex F-252 NanoTight™ Tubing Sleeve, Purple FEP, 0.042" ID x 1/16" OD; 1/EA WW-02018-09 Mfr # F-252
COMPARE
Purple 1 mm OD tubing
₹520.00INR / Each
Idex F-252X NanoTight™ Tubing Sleeve, Purple FEP, 0.042" ID x 1/16" OD; 10/PK WW-02018-10 Mfr # F-252X
COMPARE
Purple 1 mm OD tubing
₹5,000.00INR / Pkg of 10
Showing 1- 15 of 15