Idex® MicroFilter Solvent Filtration Frits
Extend filtration life and consistency with replaceable frits
  • Protects systems from damaging particulate matter
  • Provides uniform porosity for longer filtration life
  • Inexpensive replacement of frits saves time and money vs. cleaning
Easily replaceable frits save you time and money lost to frit-cleaning labor, plus eliminate the problem of embedded debris re-entering your instrument. These frits are available in the most common HPLC filtration ratings. Each includes a PCTFE or PEEK polymer sealing ring, which are slightly thicker than the frit disc for extra sealing and chemical resistance. Many also have the most common outer diameters — 0.250" and 0.254" — making them ideal for use in inline or precolumn filters.
1,900.00 - ₹33,400.00INR / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
Idex MicroFilter Frit, Stainless Steel, 2 µm, 0.094" OD, 1/4" OD PEEK Ring, 1.7 µL 0201001 Mfr # A-100
COMPARE
2.39 Stainless Steel 2
₹1,900.00INR / Each
Idex MicroFilter Frit, SS, 2 µm, 0.062" OD, 0.25" OD PEEK Ring, 0.7 µL; 1/EA 0201027 Mfr # A-101
COMPARE
1.6 Stainless Steel 2
₹1,900.00INR / Each
Idex MicroFilter Frit, SS, 0.5 µm, 0.062" OD, 0.25" OD PEEK Ring, 0.6 µL; 1/EA 0201028 Mfr # A-102
COMPARE
1.6 Stainless Steel 0.5
₹1,900.00INR / Each
Idex MicroFilter Frit, PEEK, 2 µm, 0.062" OD, 0.25" OD PCTFE Ring, 0.7 µL; 1/EA 0201036 Mfr # A-700
COMPARE
2.4 PEEK 2
₹3,500.00INR / Each
Idex MicroFilter Frit, PEEK, 0.5 µm, 0.062" OD, 0.25" OD PCTFE Ring, 0.6 µL; 1/EA 0201037 Mfr # A-701
COMPARE
1.57 PEEK 0.5
₹3,500.00INR / Each
Idex MicroFilter Frit, PEEK/PCTFE, 0.5 µm, 0.045" OD Frit, 0.192" OD Ring; 10/PK 0201043 Mfr # A-735X
COMPARE
1.143 PEEK/PCTFE 0.5
₹33,400.00INR / Pkg of 10
Idex MicroFilter Frit, SS/PEEK, 2 µm, 0.038" OD, 0.192" OD Ring; 1/EA 0201044 Mfr # C-420
COMPARE
0.97 Stainless Steel 2
₹3,700.00INR / Each
Idex MicroFilter Frit, SS, 0.5 µm, 0.094" OD, 0.25" OD PEEK Ring, 1.4 µL; 1/EA 0201054 Mfr # A-103
COMPARE
2.39 Stainless Steel 0.5
₹1,900.00INR / Each
Idex MicroFilter Frit, SS, 2 µm, 0.125" OD, 0.20" OD PCTFE Ring; 1/EA 0201064 Mfr # A-420
COMPARE
3.18 Stainless Steel 2
₹3,000.00INR / Each
Idex MicroFilter Frit, PEEK, 2 µm, 0.091" OD, 0.25" OD PCTFE Ring; 1/EA 0201065 Mfr # A-702
COMPARE
2.31 PEEK 2
₹3,600.00INR / Each
Idex MicroFilter Frit, PEEK, 0.5 µm, 0.092" OD, 0.25" OD PCTFE Ring; 1/EA 0201066 Mfr # A-703
COMPARE
2.34 PEEK 0.5
₹3,500.00INR / Each
Idex MicroFilter Frit, PEEK, 2 µm, 0.125" OD, 0.25" OD PCTFE Ring; 1/EA 0201067 Mfr # A-704
COMPARE
3.18 PEEK 2
₹3,600.00INR / Each
Idex MicroFilter Frit, PEEK, 2 µm, 0.188" OD, 0.25" OD PCTFE Ring, 7.1 µL; 1/EA 0201068 Mfr # A-706
COMPARE
4.8 PEEK 2
₹3,500.00INR / Each
Idex MicroFilter Frit, PEEK, 0.5 µm, 0.195" OD, 0.25" OD PCTFE Ring, 6.1 µL; 1/EA 0201069 Mfr # A-707
COMPARE
4.95 PEEK 0.5
₹3,500.00INR / Each
Idex MicroFilter Frit, PEEK, 2 µm, 0.062" OD, 0.20" OD PCTFE Ring; 1/EA 0201070 Mfr # A-708
COMPARE
1.57 PEEK 2
₹4,200.00INR / Each
Idex MicroFilter Frit, PEEK, 2 µm, 0.125" OD, 0.20" OD PCTFE Ring; 1/EA 0201071 Mfr # A-710
COMPARE
3.18 PEEK 2
₹3,600.00INR / Each
Idex MicroFilter Frit, SS, 0.5 µm, 0.038" OD, 0.125" OD PEEK Ring; 1/EA 0201074 Mfr # C-128-31
COMPARE
0.97 Stainless Steel 0.5
₹2,800.00INR / Each
Idex MicroFilter Frit, Stainless Steel, 0.5 µm, 0.188" OD, 0.254" OD PCTFE Ring 0201075 Mfr # C-140-30
COMPARE
4.8 Stainless Steel 0.5
₹4,200.00INR / Each
Idex MicroFilter Frit, SS, 2 µm, 0.125" OD, 0.25" OD PEEK Ring; 1/EA 0201076 Mfr # C-401
COMPARE
3.18 Stainless Steel 2
₹1,900.00INR / Each
Idex MicroFilter Frit, SS, 2 µm, 0.188" OD, 0.254" OD PEEK Ring; 1/EA 0201077 Mfr # C-402
COMPARE
4.8 Stainless Steel 2
₹2,000.00INR / Each
Showing 1 - 20 of 24