Idex® MarvelXACT Torque-Limiting Stainless Steel UHPLC Fitting Assemblies
Use MarvelXACT UHPLC connections for trouble-free performance
  • Finger-tight seals up to 19,000 psi, without the chance of under- or over-tightening
  • Durable — allows more than 100 connects and disconnects
  • No peak tailing or broadening
  • Seals at the bottom of the port for zero dead volume
  • Compatible with 10-32 coned receiving ports
MarvelXACT stainless steel fittings ensure a perfect connection every time and can be used for more than 100 connects and disconnects, providing trouble-free liquid chromatography. The connection system is small but accessible, allowing finger-tightening with a "click" that ensures optimal torque. Proprietary sealing technology means zero extra internal volume.
21,100.00 - ₹30,900.00INR / Each
27 variations of this product are available.
See All Product Options Below
27 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Fitting Length (mm)
Availability Pricing
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 100 µm ID x 150 mm Length Kit WW-42701-86 Mfr # UPFS-7100150
COMPARE
PEEK/SS 150
₹26,900.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 100 µm ID x 350 mm Length Kit WW-42701-88 Mfr # UPFS-7100350
COMPARE
PEEK/SS 350
₹27,200.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 100 µm ID x 500 mm Length Kit WW-42701-89 Mfr # UPFS-7100500
COMPARE
PEEK/SS 500
₹28,200.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 125 µm ID x 150 mm Length Kit WW-42701-91 Mfr # UPFS-7125150
COMPARE
PEEK/SS 150
₹23,100.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 125 µm ID x 250 mm Length Kit WW-42701-92 Mfr # UPFS-7125250
COMPARE
PEEK/SS 250
₹23,100.00INR / Each
Idex MarvelXACT Stainless Steel High-Pressure Connection System; 125 µm ID x 350 mm Length Kit WW-42701-93 Mfr # UPFS-7125350
COMPARE
PEEK/SS 350
₹23,100.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 125 µm ID x 500 mm Length Kit WW-42701-94 Mfr # UPFS-7125500
COMPARE
PEEK/SS 500
₹24,200.00INR / Each
Idex MarvelXACT Stainless Steel High-Pressure Connection System; 125 µm ID x 600 mm Length Kit WW-42701-95 Mfr # UPFS-7125600
COMPARE
PEEK/SS 600
₹24,200.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 254 µm ID x 150 mm Length Kit WW-42701-96 Mfr # UPFS-7254150
COMPARE
PEEK/SS 150
₹21,100.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 254 µm ID x 250 mm Length Kit WW-42701-97 Mfr # UPFS-7254250
COMPARE
PEEK/SS 250
₹22,100.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 254 µm ID x 500 mm Length Kit WW-42701-99 Mfr # UPFS-7254500
COMPARE
PEEK/SS 500
₹24,500.00INR / Each
Idex MarvelXACT MarvelXACT Stainless Steel High-Pressure Connection System; 254 µm ID x 600 mm Length Kit WW-42702-01 Mfr # UPFS-7254600
COMPARE
PEEK/SS 600
₹23,400.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 25 µm ID x 150 mm Length Kit WW-42702-02 Mfr # UPFP-7025150
COMPARE
PEEK/SS 150
₹28,900.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 25 µm ID x 250 mm Length Kit WW-42702-03 Mfr # UPFP-7025250
COMPARE
PEEK/SS 250
₹28,900.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 25 µm ID x 350 mm Length Kit WW-42702-04 Mfr # UPFP-7025350
COMPARE
PEEK/SS 350
₹29,900.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 25 µm ID x 500 mm Length Kit WW-42702-05 Mfr # UPFP-7025500
COMPARE
PEEK/SS 500
₹29,900.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 50 µm ID x 600 mm Length Kit WW-42702-11 Mfr # UPFP-7050600
COMPARE
PEEK/SS 600
₹30,900.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 75 µm ID x 150 mm Length Kit WW-42702-12 Mfr # UPFP-7075150
COMPARE
PEEK/SS 150
₹28,900.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 75 µm ID x 250 mm Length Kit WW-42702-13 Mfr # UPFP-7075250
COMPARE
PEEK/SS 250
₹28,900.00INR / Each
Idex MarvelXACT PEEK-Lined Stainless Steel High-Pressure Connection System; 75 µm ID x 350 mm Length Kit WW-42702-14 Mfr # UPFP-7075350
COMPARE
PEEK/SS 350
₹29,900.00INR / Each
Showing 1 - 20 of 27