Idex® MarvelX Stainless Steel UHPLC Connections
Advance your fluidic systems with MarvelX SS Fittings
  • Finger-tight seals up to 19,000 PSI for routine use
  • Flexible tubing is replaceable independent of fittings, reducing waste
  • No peak tailing
  • High plate efficiency
MarvelX convenient, removable stainless steel fittings can be used for more than 100 connects and disconnects, providing easy routing and consistent performance throughout your instrument. The connection system is small but accessible, allowing finger tightening that eliminates the need to use tools in narrow spaces. Flexible tubing can be replaced independent of fittings, providing superior reusability and reducing waste. Choose PEEK-lined assemblies for your bio-inert applications.
21,300.00 - ₹31,200.00INR / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Fitting Length (mm) Description
Availability Pricing
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 125 µm ID x 150 mm Length Kit WW-42701-01 Mfr # UPFS-6125150
COMPARE
Stainless Steel 150 Stainless Steel High-Pressure Connection System; 125 µm ID x 150 mm Length Kit
₹23,800.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 254 µm ID x 150 mm Length Kit WW-42701-06 Mfr # UPFS-6254150
COMPARE
Stainless Steel 150 Stainless Steel High-Pressure Connection System; 254 µm ID x 150 mm Length Kit
₹21,800.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 254 µm ID x 350 mm Length Kit WW-42701-08 Mfr # UPFS-6254350
COMPARE
Stainless Steel 350 Stainless Steel High-Pressure Connection System; 254 µm ID x 350 mm Length Kit
₹22,400.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 100 µm ID x 350 mm Length Kit WW-42701-16 Mfr # UPFS-6100350
COMPARE
Stainless Steel 350 Stainless Steel High-Pressure Connection System; 100 µm ID x 350 mm Length Kit
₹27,400.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 100 µm ID x 500 mm Length Kit WW-42701-18 Mfr # UPFS-6100500
COMPARE
Stainless Steel 500 Stainless Steel High-Pressure Connection System; 100 µm ID x 500 mm Length Kit
₹28,400.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 100 µm ID x 600 mm Length Kit WW-42701-20 Mfr # UPFS-6100600
COMPARE
Stainless Steel 600 Stainless Steel High-Pressure Connection System; 100 µm ID x 600 mm Length Kit
₹28,400.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 125 µm ID x 250 mm Length Kit WW-42701-24 Mfr # UPFS-6125250
COMPARE
Stainless Steel 250 Stainless Steel High-Pressure Connection System; 125 µm ID x 250 mm Length Kit
₹23,400.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 125 µm ID x 500 mm Length Kit WW-42701-26 Mfr # UPFS-6125500
COMPARE
Stainless Steel 500 Stainless Steel High-Pressure Connection System; 125 µm ID x 500 mm Length Kit
₹24,400.00INR / Each
Idex MarvelX Stainless Steel High-Pressure Connection System; 254 µm ID x 70 mm Length Kit WW-42701-30 Mfr # UPFS-6254070
COMPARE
Stainless Steel 70 Stainless Steel High-Pressure Connection System; 254 µm ID x 70 mm Length Kit
₹21,300.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 254 µm ID x 250 mm Length Kit WW-42701-32 Mfr # UPFS-6254250
COMPARE
Stainless Steel 250 Stainless Steel High-Pressure Connection System; 254 µm ID x 250 mm Length Kit
₹22,800.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 254 µm ID x 500 mm Length Kit WW-42701-34 Mfr # UPFS-6254500
COMPARE
Stainless Steel 500 Stainless Steel High-Pressure Connection System; 254 µm ID x 500 mm Length Kit
₹23,400.00INR / Each
Idex MarvelX MarvelX Stainless Steel High-Pressure Connection System; 254 µm ID x 600 mm Length Kit WW-42701-36 Mfr # UPFS-6254600
COMPARE
Stainless Steel 600 Stainless Steel High-Pressure Connection System; 254 µm ID x 600 mm Length Kit
₹23,400.00INR / Each
Idex MarvelX PEEK-Lined Stainless Steel High-Pressure Connection System; 100 µm ID x 150 mm Length Kit WW-42701-40 Mfr # UPFP-6100150
COMPARE
Stainless Steel 150 Stainless Steel High-Pressure Connection System; 100 µm ID x 150 mm Length Kit
₹29,200.00INR / Each
Idex MarvelX MarvelX PEEK-Lined Stainless Steel High-Pressure Connection System; 100 µm ID x 350 mm Length Kit WW-42701-44 Mfr # UPFP-6100350
COMPARE
Stainless Steel 350 Stainless Steel High-Pressure Connection System; 100 µm ID x 350 mm Length Kit
₹30,200.00INR / Each
Idex MarvelX PEEK-Lined Stainless Steel High-Pressure Connection System; 100 µm ID x 70 mm Length Kit WW-42701-52 Mfr # UPFP-6100070
COMPARE
Stainless Steel 70 Stainless Steel High-Pressure Connection System; 100 µm ID x 70 mm Length Kit
₹29,200.00INR / Each
Idex MarvelX MarvelX PEEK-Lined Stainless Steel High-Pressure Connection System; 100 µm ID x 250 mm Length Kit WW-42701-54 Mfr # UPFP-6100250
COMPARE
Stainless Steel 250 Stainless Steel High-Pressure Connection System; 100 µm ID x 250 mm Length Kit
₹29,200.00INR / Each
Idex MarvelX MarvelX PEEK-Lined Stainless Steel High-Pressure Connection System; 100 µm ID x 500 mm Length Kit WW-42701-56 Mfr # UPFP-6100500
COMPARE
Stainless Steel 500 Stainless Steel High-Pressure Connection System; 100 µm ID x 500 mm Length Kit
₹30,200.00INR / Each
Idex MarvelX MarvelX PEEK-Lined Stainless Steel High-Pressure Connection System; 100 µm ID x 600 mm Length Kit WW-42701-58 Mfr # UPFP-6100600
COMPARE
Stainless Steel 600 Stainless Steel High-Pressure Connection System; 100 µm ID x 600 mm Length Kit
₹31,200.00INR / Each
Idex MarvelX Stainless Steel High-Pressure Connection System; 50 µm ID x 70 mm Length Kit WW-42701-62 Mfr # UPFP-6050070
COMPARE
Stainless Steel 70 Stainless Steel High-Pressure Connection System; 50 µm ID x 70 mm Length Kit
₹29,200.00INR / Each
Idex MarvelX Stainless Steel High-Pressure Connection System; 75 µm ID x 70 mm Length Kit WW-42701-74 Mfr # UPFP-6075070
COMPARE
Stainless Steel 70 Stainless Steel High-Pressure Connection System; 75 µm ID x 70 mm Length Kit
₹29,200.00INR / Each
Showing 1 - 20 of 21