Idex® Low-Pressure Crosses
Combine, split, or intersect fluid streams throughout your system
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
Use crosses in a variety of low-pressure situations throughout your fluidic system. Select assemblies that come complete with nuts and ferrules or order bodies and provide your own fittings. Each cross comes with handy mounting holes.
9,300.00 - ₹15,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 2) (in) Fitting Material Description
Availability Pricing
Idex Low-Pressure Cross Assembly, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0200820 Mfr # P-722
COMPARE
1/16 PEEK Low-Pressure Cross Assembly, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹15,000.00INR / Each
Idex Low-Pressure Cross Assembly, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0200824 Mfr # P-723
COMPARE
1/8 PEEK Low-Pressure Cross Assembly, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹15,000.00INR / Each
Idex Low-Pressure Cross Assembly, Natural ETFE, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202350 Mfr # P-634
COMPARE
1/16 ETFE Low-Pressure Cross Assembly, Natural ETFE, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹13,100.00INR / Each
Idex Low-Pressure Cross Body, Natural ETFE, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202351 Mfr # P-634-01
COMPARE
1/16 ETFE Low-Pressure Cross Body, Natural ETFE, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹10,100.00INR / Each
Idex Low-Pressure Cross Assembly, Natural ETFE, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202352 Mfr # P-635
COMPARE
1/8 ETFE Low-Pressure Cross Assembly, Natural ETFE, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹13,100.00INR / Each
Idex Low-Pressure Cross Body, Natural ETFE, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202353 Mfr # P-635-01
COMPARE
1/8 ETFE Low-Pressure Cross Body, Natural ETFE, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹9,700.00INR / Each
Idex Low-Pressure Cross Body, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202379 Mfr # P-722-01
COMPARE
1/16 PEEK Low-Pressure Cross Body, Natural PEEK, 0.020" Bore, 1/16" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹9,300.00INR / Each
Idex Low-Pressure Cross Body, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202380 Mfr # P-723-01
COMPARE
1/8 PEEK Low-Pressure Cross Body, Natural PEEK, 0.050" Bore, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
₹9,500.00INR / Each
Showing 1- 8 of 8