Idex® LiteTouch® Chromatography Fittings, 1/4-28 Coned Connection for 1/8" OD Tubing
High pressure fittings with fingertight convenience
  • Grips tubing at two pressure points
  • Design prevents tubing from twisting
Combine a LiteTouch® nut and LiteTouch ferrule to create your fitting assembly. The LiteTouch fitting assembly grips tubing at two compression points allowing fittings to withstand high pressures with fingertight convenience. Ferrule with lock ring prevents polymer tubing from twisting. Use with hard tubing, such as stainless steel and PEEK tubing.
1,200.00 - ₹11,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex LifeTouch® FlushNut™, 316 SS, 1/8" OD Tubing, 1/4-28 Coned; 1/EA 0202111 Mfr # F-364
COMPARE
Natural Stainless steel LifeTouch® FlushNut™, 316 SS, 1/8" OD Tubing, 1/4-28 Coned; 1/EA
₹4,500.00INR / Each
Idex LiteTouch® Nut, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Coned; 1/EA 0202243 Mfr # LT-215
COMPARE
Natural PEEK LiteTouch® Nut, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Coned; 1/EA
₹1,200.00INR / Each
Idex LiteTouch® Nut, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Coned; 10/PK 0202248 Mfr # LT-215X
COMPARE
Natural PEEK LiteTouch® Nut, Short, Natural PEEK, 1/8" OD Tubing, 1/4-28 Coned; 10/PK
₹11,200.00INR / Pkg of 10