Idex® High-Pressure UNF to NPT(M) Threaded Adapters
Maintain secure connections to 1/8" and 1/4" NPT ports even under high pressure
  • Securely join together fluid pathway components
  • Manufactured from PEEK polymer
Made from PEEK, these adapters are durable and have excellent chemical resistance. Wrap NPT threads with thread seal tape to ensure leak-free seal.
3,900.00 - ₹9,300.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Thread size UNF(F) (port 1) (InchShort) Thread size UNF(M) (port 1) (InchShort) Thread Size NPT(M) (Port 2) (InchShort)
Availability Pricing
Idex Threaded Adapter, Red PEEK, 0.062" ID, 1/4-28 Flat Bottom (F) to 1/4" NPT(M) 0202494 Mfr # U-500-01
COMPARE
1/4-28 Flat Bottom
₹3,900.00INR / Each
Idex Threaded Adapter, Red PEEK, 0.062" ID, 1/4-28 Flat Bottom (M) to 1/4" NPT(M) 0202495 Mfr # U-501
COMPARE
1/4-28 Flat Bottom
₹9,300.00INR / Each
Idex Threaded Adapter, PEEK, 1/8" ID, 5/16-24 Flat Bottom (F) to 1/4" NPT(M) 0202500 Mfr # U-504-01
COMPARE
5/16-24 Flat Bottom
₹6,000.00INR / Each
Idex Threaded Adapter, Red PEEK, 0.062" ID, 1/4-28 Flat Bottom (F) to 1/8" NPT(M) 0202501 Mfr # U-510-01
COMPARE
1/4-28 Flat Bottom
₹4,000.00INR / Each
Idex Threaded Adapter, Red PEEK, 0.062" ID, 1/4-28 Flat Bottom (M) to 1/8" NPT(M) 0202502 Mfr # U-511
COMPARE
1/4-28 Flat Bottom
₹8,500.00INR / Each
Idex Threaded Adapter, PEEK, 1/8" ID, 5/16-24 Flat Bottom (F) to 1/8" NPT(M) 0202504 Mfr # U-514-01
COMPARE
5/16-24 Flat Bottom
₹4,000.00INR / Each