Idex® High-Pressure UNF to M6 (F) Adapters
Maintain secure connections even under high pressure
  • Securely join together fluid pathway components
  • Manufactured from inert polymers
  • Holds tight connection even through vibration
Made from PEEK or PCTFE, these adapters are durable and have excellent chemical resistance.
5,700.00 - ₹12,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Thread size UNF(M) (port 1) (InchShort) Orifice Dimensions (in)
Availability Pricing
Idex Threaded Adapter, PEEK, 0.030" ID, 10-32 Coned (M) to M6 Flat Bottom (F) 0201410 Mfr # P-650
COMPARE
PEEK 10-32 Coned 1/32
₹5,700.00INR / Each
Idex Threaded Adapter with Fitting, PEEK, 0.020" ID, 10-32 Coned (F) to M6 Flat Bottom (F) 0202435 Mfr # P-626
COMPARE
PEEK 10-32 Coned 1/50
₹12,700.00INR / Each
Idex Threaded Adapter, PEEK, 0.030" ID, 1/4-28 Flat Bottom (M) to M6 Flat Bottom (F) 0202441 Mfr # P-670
COMPARE
PEEK 1/4-28 Flat Bottom 1/32
₹8,700.00INR / Each