Idex® High-Pressure UNF(F) to M6 (M) Adapters
Maintain secure connections even under high pressure
  • Securely join together fluid pathway components
  • Manufactured from inert polymers
  • Holds tight connection even through vibration
Made from PEEK or PCTFE, these adapters are durable and have excellent chemical resistance.
4,900.00 - ₹5,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Thread Size ISO Metric (M) (Port 2) (mm) Fitting Material Thread size UNF(F) (port 1) (in)
Availability Pricing
Idex Threaded Adapter, PEEK, 0.040" ID, 5/16-24 Flat Bottom (F) to M6 Flat Bottom (M) WW-02024-44 Mfr # P-673
COMPARE
6 PEEK 5/16-24 Flat Bottom
₹5,900.00INR / Each
Idex Threaded Adapter, PEEK, 0.040" ID, 1/4-28 Flat Bottom (F) to M6 Flat Bottom (M) WW-02024-48 Mfr # P-694
COMPARE
6 PEEK 1/4-28 Flat Bottom
₹5,400.00INR / Each
Idex Threaded Adapter, PEEK, 0.040" ID, 10-32 Coned (F) to M6 Flat Bottom (M) WW-02024-80 Mfr # P-920-01
COMPARE
6 PEEK 10-32 Coned
₹4,900.00INR / Each