Idex® High-Pressure MicroTight® Unions and Elbow
Securely connect two-pieces of capillary tubing with MicroTight connectors
  • Unions withstand up to 5000 psi (345 bar)
Use the unions and elbows to connect two pieces of capillary tubing that will withstand higher pressures. The true zero-dead-volume (ZDV) unions feature a gauge plug that allows two pieces of tubing to connect directly to each other to ensure proper alignment. Use the conductive MicroTight union to introduce voltage into your system.
4,400.00 - ₹18,500.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Thread size UNF(F) (port 1) (in)
Availability Pricing
Idex High-Pressure MicroTight Union Assembly, True ZDV, PEEK, 1/32" OD Tubing, 6-32 Coned; 1/EA WW-02014-43 Mfr # P-771
COMPARE
1/32 PEEK 6-32 Coned
₹7,700.00INR / Each
Idex High-Pressure MicroTight Union Assembly, MicroTight, PEEK, 0.006" ID, 360 µm OD Tubing, 5/16-24 Coned; 1/EA WW-02014-44 Mfr # P-772
COMPARE
1/64 PEEK 6-32 Coned
₹8,700.00INR / Each
Idex Conductive MicroTight® Union Assembly for 360 µm Tubing, 5/16-24 Coned WW-02024-25 Mfr # M-572
COMPARE
1/64 SS/PEEK 5/16-24 Coned
₹18,500.00INR / Each
Idex High-Pressure MicroTight Union Body, True ZDV, PEEK, 1/32" OD Tubing, 6-32 Coned; 1/EA WW-02024-61 Mfr # P-771-01
COMPARE
1/32 PEEK 6-32 Coned
₹4,400.00INR / Each