Idex® High-Pressure MicroTight® Adapters for 1/16" OD to 1/32" OD
Create a high-pressure connection between 1/16" OD tubing and capillary tubing
  • Securely join together fluid pathway components
  • Manufactured from 316 stainless steel or inert polymers
  • Holds tight connection even through vibration
Port 1 is 10-32 coned for 1/16" OD tubing.
13,900.00 - ₹28,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Thread size UNF(F) (port 2) (InchShort) Description
Availability Pricing
Idex MicroTight Adapter Assy, True ZDV, Red PEEK, 1/16 to 1/32" OD, 10-32 to 6-32 0202468 Mfr # P-881
COMPARE
PEEK 6-32 Coned MicroTight Adapter Assembly, True ZDV, Red PEEK, 1/16 to 1/32" OD Tubing, 10-32 Coned to 6-32 Coned; 1/EA
₹13,900.00INR / Each
Idex Ultra High-Pressure MicroAdapter Assy, SS, 1/16" to 1/32" OD, 10-32 to 6-32 0202509 Mfr # UH-630
COMPARE
SS/PEEK 6-32 Coned UltraHigh-Pressure MicroAdapter Assembly, SS, 1/16" to 1/32" OD Tubing, 10-32 to 6-32 Coned; 1/EA
₹28,600.00INR / Each