IDEX® Fused Silica Chromatography Tubing
Great combination of quality, purity, strength, and flexibility
  • Tight, ultra-smooth interior surface is ideal for micro-scale analyses
  • Well suited for analytical sciences
Fused silica tubing is made from high-quality, polyimide-clad fused silica. It offers excellent flexibility, tensile strength, and abrasion resistance. Use tubing cutters to ensure precise, clean cuts of fused silica tubing.
37,600.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (m) Tubing ID (mm) Tubing OD (mm)
Availability Pricing
Idex FS-110 Fused Silica Tubing, 360 OD x 100 ID x 2 m 0203232 Mfr # FS-110
COMPARE
2 0.1 0.36
₹37,600.00INR / Each
Idex FS-115 Fused Silica Tubing, 360 OD x 150 ID x 2 m 0203233 Mfr # FS-115
COMPARE
2 0.15 0.36
₹37,600.00INR / Each
Idex FS-120 Fused Silica Tubing, 360 OD x 20 ID x 2 m 0203234 Mfr # FS-120
COMPARE
2 0.02 0.36
₹37,600.00INR / Each
Idex FS-150 Fused Silica Tubing, 360 OD x 50 ID x 2 m 0203235 Mfr # FS-150
COMPARE
2 0.05 0.36
₹37,600.00INR / Each
Idex FS-175 Fused Silica Tubing, 360 OD x 75 ID x 2m 0203236 Mfr # FS-175
COMPARE
2 0.075 0.36
₹37,600.00INR / Each